Skip to Content
Merck
All Photos(1)

Key Documents

242861

Sigma-Aldrich

Chloroiodomethane

97%

Synonym(s):

Chloromethyl iodide, Iodochloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2I
CAS Number:
Molecular Weight:
176.38
Beilstein:
1730802
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.582 (lit.)

bp

108-109 °C (lit.)

density

2.422 g/mL at 25 °C (lit.)

functional group

chloro
iodo

SMILES string

ClCI

InChI

1S/CH2ClI/c2-1-3/h1H2

InChI key

PJGJQVRXEUVAFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photolysis of chloroiodomethane in cryogenic matrices along with recombination of the nascent radical pair was studied.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Sumner et al.
Environmental science & technology, 53(14), 8216-8226 (2019-07-06)
A number of flowback samples derived from horizontally drilled hydraulic fracturing (HDHF) operations reveal consistent detections of halogenated organic species , yet the source of these compounds remains uncertain. Studies simulating subsurface conditions have found that oxidative "breakers" can halogenate certain
Thomas J Preston et al.
The Journal of chemical physics, 135(11), 114503-114503 (2011-09-29)
Photolysis of chloroiodomethane (CH(2)ClI) in cryogenic matrices followed by recombination of the nascent radical pair produces an isomer (CH(2)Cl-I) that features a halogen-halogen (Cl-I) bond. Using ultrafast laser pulses, it is possible to follow the formation of this isomer by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service