Skip to Content
Merck
All Photos(3)

Key Documents

244961

Sigma-Aldrich

Tetrakis(triphenylphosphine)platinum(0)

97%

Synonym(s):

Pt(PPh3)4, Platinum(0)-tetrakis(triphenylphosphine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt[(C6H5)3P]4
CAS Number:
Molecular Weight:
1244.22
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: platinum
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[Pt].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Pt/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

SYKXNRFLNZUGAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrakis(triphenylphosphine)platinum(0) is used as a precursor to platinum complexes.
It can undergo oxidative addition to organochalcogen compounds such as dithienyl ditelluride through the cleavage of the carbon–chalcogen bond. This reaction can play a role in homogeneous catalysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The X-ray crystallographic study of the reaction of bis (2-thienyl) ditelluride with tetrakis (triphenylphosphine) platinum or-palladium.
Oilunkaniemi R, et al.
Journal of Organometallic Chemistry, 595(2), 232-240 (2000)
New oxidative substitution reactions of tetrakis (triphenylphosphine) platinum (0).
Tayim H A and Akl N S
J. Inorg. Nucl. Chem., 36(5), 1071-1074 (1974)
Zerovalent platinum chemistry. VIII. The reactions of nitric oxide with tris-and tetrakis (triphenylphosphine) platinum (0).
Cenini S, et al.
Inorgorganica Chimica Acta, 6, 182-184 (1972)
Reactions of elemental sulfur with tetrakis (triphenylphosphine) platinum (0). Formation of a complex containing very nucleophilic bridging sulfido ligands.
Gukathasan R R, et al.
Canadian Journal of Chemistry, 61(11), 2490-2492 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service