Skip to Content
Merck
All Photos(1)

Documents

220809

Sigma-Aldrich

Diethylzinc solution

15 wt. % in toluene

Synonym(s):

Zincdiethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2Zn
CAS Number:
Molecular Weight:
123.51
Beilstein:
3587207
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

15 wt. % in toluene

density

0.915 g/mL at 25 °C

SMILES string

CC[Zn]CC

InChI

1S/2C2H5.Zn/c2*1-2;/h2*1H2,2H3;

InChI key

HQWPLXHWEZZGKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed together with CF3I in a Rh(I) catalyzed preparation of α-trifluoromethyl ketones from α,ß-unsaturated ketones. Promoter for the Wittig reaction of carbonyl compounds with dibromofluoroacetate and triphenylphosphine leading to α-fluroroacrylates. Ni catalyzed coupling with cyclic anhydrides.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkene-directed regioselective nickel-catalyzed cross-coupling of cyclic anhydrides with diorganozinc reagents.
Rebecca L Rogers et al.
Angewandte Chemie (International ed. in English), 46(48), 9301-9304 (2007-11-07)
Synthesis, 3409-3409 (2006)
Organic Syntheses, 83, 177-177 (2006)
Zhiyan Huang et al.
The Journal of organic chemistry, 72(4), 1373-1378 (2007-02-10)
A novel class of sulfinamido alcohol ligands 1-6 was synthesized from (S)-tert-butanesulfinamide. These ligands showed excellent catalytic activities and enantiomeric selectivities in asymmetric additions of diethylzinc to aromatic aldehydes.
Weimin Lin et al.
The Journal of organic chemistry, 74(2), 645-651 (2008-12-06)
The application of a zinc carbenoid-mediated chain-extension reaction to a functionalized peptide isostere is reported. The cleavage site of human CVM protease was utilized as a target for testing the synthetic methodology. The utility of this chain-extension reaction is demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service