Skip to Content
Merck
All Photos(3)

Key Documents

241547

Sigma-Aldrich

Lead oxide

powder (red), 1-2 μm, 99%

Synonym(s):

Lead oxide, red, Lead(II,IV) oxide, Minium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb3O4
CAS Number:
Molecular Weight:
685.60
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

10 mmHg ( 0 °C)

Assay

99%

form

powder (red)

particle size

1-2 μm

SMILES string

O1[PbH2]O[Pb]12O[PbH2]O2

InChI

1S/4O.3Pb

InChI key

XMFOQHDPRMAJNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Average particle size determined by Fisher sub-sieve sizer

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Ox. Sol. 2 - Repr. 1A - STOT RE 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elżbieta Horszczaruk et al.
Materials (Basel, Switzerland), 13(21) (2020-11-05)
In recent years, a nano-modification of the cement composites allowed to develop a number of new materials. The use of even small amount of nano-admixture makes possible not only to improve the physico-mechanical properties of the cement materials, but also
Lead intoxication in children.
W L Nyhan et al.
The Western journal of medicine, 143(3), 357-364 (1985-09-01)
Shujun Zhang et al.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control, 52(11), 2131-2139 (2006-01-21)
The perovskite solid solution system (1-x)BiScO3-(x)PbTiO3 represents an interesting new family of high-temperature piezoelectric materials. Compositions near the morphotropic phase boundary (x approximately 0.64) have been reported to have high Curie temperatures (Tc > 450 degrees C) and good piezoelectric
T Kákosy et al.
Journal of toxicology. Clinical toxicology, 34(5), 507-511 (1996-01-01)
Report of a lead intoxication epidemic caused by ingestion of contaminated ground paprika. Retrospective study of case histories. Institutional, toxicological and medical referral center for ambulatory and hospitalized care. One hundred forty-one adults consumed paprika contaminated with lead tetroxide (red
[Abnormal lead absorption due to penetration of lead tetraoxide (red-lead) in the hand (author's transl)].
F Loi
La Medicina del lavoro, 71(2), 157-160 (1980-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service