Skip to Content
Merck
All Photos(3)

Key Documents

238198

Sigma-Aldrich

Dimethyl maleate

96%

Synonym(s):

(2Z)-2-Butenedioic acid dimethyl ester, (Z)-2-Butenedioic acid dimethyl ester, (Z)-Dimethyl 2-butenedioate, Dimethyl (Z)-but-2-enedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH=CHCOOCH3
CAS Number:
Molecular Weight:
144.13
Beilstein:
471705
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

impurities

≤4% dimethyl fumarate

refractive index

n20/D 1.441 (lit.)

bp

204-205 °C (lit.)

solubility

water: soluble 77.9 g/L at 20 °C

density

1.152 g/mL at 25 °C (lit.)

SMILES string

[H]\C(=C(/[H])C(=O)OC)C(=O)OC

InChI

1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3-

InChI key

LDCRTTXIJACKKU-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl maleate is a dienophile used as a chemical intermediate to synthesize dienes, plastics, and copolymers.

Dimethyl maleate (DMM) is a reactive dienophile and undergoes ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. Mesoporous siliceous SBA-15-supported Cu catalyzed gas phase hydrogenolysis of DMM to 1,4-butanediol (BDO) has been reported. Aluminium chloride has been reported to accelerate the Diels-Alder reaction of DMM and anthracene. DMM can be synthesized by the esterification of maleic anhydride with sulfuric acid and methanol.

Application

  • Dissociation of bovine 6S procarboxypeptidase A by reversible condensation with 2,3-dimethyl maleic anhydride: application to the partial characterization of subunit III.: This study explores the dissociation of bovine procarboxypeptidase A using 2,3-dimethyl maleic anhydride, highlighting its applications in the partial characterization of enzyme subunits. The research demonstrates the potential of dimethyl maleate derivatives in protein chemistry and enzyme structure studies. (Puigserver and Desnuelle, 1975).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT RE 2 Dermal - STOT SE 3

Target Organs

Respiratory system, Skin

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acceleration of the Diels-Alder reaction by aluminum chloride.
Yates P and Eaton P.
Journal of the American Chemical Society, 82(16), 4436-4437 (2960)
James M Longmire et al.
Journal of the American Chemical Society, 124(45), 13400-13401 (2002-11-07)
A highly reactive Ag(I)-catalyzed [3 + 2] cycloaddition of azomethine ylides is founded using AgOAc as the catalytic precursor and phosphines as ligands. Using a new bis-ferrocenyl amide phosphine (FAP) as the ligand, we found that high enantioselectivities (up to
Mitsuo Miyazawa et al.
Natural product research, 17(5), 337-339 (2003-10-07)
Dimethyl malate (1) and 5-hydroxymethyl furfural (2) were isolated as insecticidal compounds by bioassay-guided fractionation from MeOH extract of the fruits of Cornus officinalis Sieb. et Zucc. Insecticidal activity against larvae of D. melanogaster was demonstrated: 1 and 2 gave
Danhong Lu et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(5), 2708-2713 (2002-03-07)
Cellular metabolism of glucose is required for stimulation of insulin secretion from pancreatic beta cells, but the precise metabolic coupling factors involved in this process are not known. In an effort to better understand mechanisms of fuel-mediated insulin secretion, we
Kun Wei et al.
The Journal of organic chemistry, 69(17), 5763-5765 (2004-08-17)
Ultrasonic irradiation effectively promotes the Diels-Alder reaction of substituted furans with reactive dienophiles (i.e., dimethyl acetylenedicarboxylate (DMAD) and dimethyl maleate). Regiospecific furano Diels-Alder cycloaddition of 2-vinylic furans with DMAD furnished functionalized oxabicyclic alkenes in good yield under ultrasonication condition.

Articles

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service