Skip to Content
Merck
All Photos(1)

Key Documents

374318

Sigma-Aldrich

Dimethyl (S)-(−)-malate

98%

Synonym(s):

Dimethyl (S)-2-hydroxysuccinate, Dimethyl L-malate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCH2CH(OH)CO2CH3
CAS Number:
Molecular Weight:
162.14
Beilstein:
1724363
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −6.5°, neat

refractive index

n20/D 1.435 (lit.)

density

1.223 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)C[C@H](O)C(=O)OC

InChI

1S/C6H10O5/c1-10-5(8)3-4(7)6(9)11-2/h4,7H,3H2,1-2H3/t4-/m0/s1

InChI key

YSEKNCXYRGKTBJ-BYPYZUCNSA-N

Application

Dimethyl (S)-(-)-malate may be used as a starting material to synthesize (-)-tulipalin B. The selective reduction of its ester group to alcohol can be accomplished using borane-dimethyl sulfide complex (BMS) in the presence of sodium tetrahydroborate.
This chiral synthon has been used to prepare cytochrome P450 metabolites of arachidonic acid, and cyclic sulfolanes with HIV-1 protease inhibition potential.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F

Flash Point(C)

55 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Falck, J.R. et al.
Tetrahedron Letters, 33, 4893-4893 (1992)
Use of enzymic hydrolysis of dimethyl malates for a short synthesis of tulipalin B and of its enantiomer.
Papageorgiou C and Benezra C.
The Journal of Organic Chemistry, 50(7), 1144-1145 (1985)
Cyclic sulfolanes as novel and high affinity P2 ligands for HIV-1 protease inhibitors.
A K Ghosh et al.
Journal of medicinal chemistry, 36(7), 924-927 (1993-04-02)
Rajesh Gupta et al.
The Journal of biological chemistry, 293(47), 18086-18098 (2018-09-20)
Secreted proteins are important metabolic regulators in both healthy and disease states. Here, we sought to investigate the mechanism by which the secreted protein complement 1q-like-3 (C1ql3) regulates insulin secretion from pancreatic β-cells, a key process affecting whole-body glucose metabolism.
Combination of borane-dimethyl sulfide complex with catalytic sodium tetrahydroborate as a selective reducing agent for a-hydroxy esters. Versatile chiral building block from (s)-(-)-malic acid.
Saito S, et al.
Chemistry Letters (Jpn), 13(8), 1389-1392 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service