Skip to Content
Merck
All Photos(3)

Documents

511218

Sigma-Aldrich

m-Toluidine

99%

Synonym(s):

3-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
635944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.567 (lit.)

bp

203-204 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(N)c1

InChI

1S/C7H9N/c1-6-3-2-4-7(8)5-6/h2-5H,8H2,1H3

InChI key

JJYPMNFTHPTTDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Toluidine (MT) is an aniline derivative that behaves as a fragile glass forming liquid. Its dielectric relaxation process, glass transition relaxation, relaxation modes and slow secondary relaxation have been studied. The analysis of MT crystalline phase by powder X-ray diffraction shows that it belongs to monoclinic space group, P21/c. Its structure is held together by N-H...N hydrogen bonds and van der Waals interactions.

Application

m-Toluidine may be used to form pyrrole/m-toluidine copolymers via oxidative polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The a-relaxation process in simple glass forming liquid m-toluidine. I. The temperature dependence of the dielectric response.
Cutroni M, et al.
J. Chem. Phys. , 114(16), 7118-7123 (2001)
Relaxation modes in glass forming meta-toluidine.
Carpentier LRD and Descamps M.
J. Chem. Phys. , 121(13), 6470-6477 (2004)
Ab initio structure determination of m-toluidine by powder X-ray diffraction.
Rukiah M, et al.
Journal of Applied Crystallography, 37(3), 464-471 (2004)
Andrea Mandanici et al.
The Journal of chemical physics, 122(8), 84508-84508 (2005-04-20)
The glass-former m-toluidine displays the characteristic properties of a fragile supercooled liquid, which suggest the existence of a slow secondary relaxation process. In view of the recently realized importance of such a secondary relaxation feature, we have conducted a dielectric
Synthesis and characterization of pyrrole and m-toluidine copolymers.
Li XG, et al.
Synthetic Metals, 123(3), 435-441 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service