Skip to Content
Merck
All Photos(2)

Documents

235946

Sigma-Aldrich

Tetramethylammonium iodide

99%

Synonym(s):

N,N,N-trimethyl-methanaminium iodide, TMAI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(I)
CAS Number:
Molecular Weight:
201.05
Beilstein:
3620030
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

>300 °C (lit.)

SMILES string

[I-].C[N+](C)(C)C

InChI

1S/C4H12N.HI/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

RXMRGBVLCSYIBO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Facile iodination of aromatic compounds including anilines, acids and alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdol R Hajipour et al.
The Journal of organic chemistry, 67(24), 8622-8624 (2002-11-26)
Tetramethylammonium dichloroiodate (1, TMADCI) as a mild and efficient iodination reagent was prepared. Iodination of different aromatic compounds with this reagent takes place fast and with high yields under solvent-free conditions.
Markus Welcker et al.
Genes & development, 27(23), 2531-2536 (2013-12-04)
The Fbw7 tumor suppressor targets a broad network of proteins for ubiquitylation. Here we show critical functions for Fbw7 dimerization in regulating the specificity and robustness of degradation. Dimerization enables Fbw7 to target substrates through concerted binding to two suboptimal
Marco I Ries et al.
The Journal of biological chemistry, 288(52), 37289-37295 (2013-11-15)
Metabolic profiling and structural elucidation of novel secondary metabolites obtained from derived deletion strains of the filamentous fungus Penicillium chrysogenum were used to reassign various previously ascribed synthetase genes of the roquefortine/meleagrin pathway to their corresponding products. Next to the
Jacob O Fierer et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), E1176-E1181 (2014-03-19)
Individual proteins can now often be modified with atomic precision, but there are still major obstacles to connecting proteins into larger assemblies. To direct protein assembly, ideally, peptide tags would be used, providing the minimal perturbation to protein function. However
Chi Nguyen et al.
Nature, 505(7483), 427-431 (2013-12-24)
Acyl carrier protein (ACP) transports the growing fatty acid chain between enzymatic domains of fatty acid synthase (FAS) during biosynthesis. Because FAS enzymes operate on ACP-bound acyl groups, ACP must stabilize and transport the growing lipid chain. ACPs have a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service