Skip to Content
Merck
All Photos(2)

Documents

374350

Sigma-Aldrich

Tridodecylmethylammonium iodide

97%

Synonym(s):

Methyltridodecylammonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]3N(I)CH3
CAS Number:
Molecular Weight:
663.93
Beilstein:
4260774
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

106-107 °C (lit.)

SMILES string

[I-].CCCCCCCCCCCC[N+](C)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C37H78N.HI/c1-5-8-11-14-17-20-23-26-29-32-35-38(4,36-33-30-27-24-21-18-15-12-9-6-2)37-34-31-28-25-22-19-16-13-10-7-3;/h5-37H2,1-4H3;1H/q+1;/p-1

InChI key

JRLISGAUAJZAGL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tridodecylmethylammonium iodide (3C12MAI) can be used:
  • As a triple-chained cationic surfactant.
  • to prepapre glucose oxidase enzyme immobilized iodide electrode for the detection of glucose concentration.
  • As a cationic phase-transfer catalyst in the oxidation of hydrocarbons by O2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of onium salts in the oxidation of hydrocarbons by O 2 catalysed by cationic phase-transfer reagents
Csanyi L, et al.
Physical Chemistry Chemical Physics, 2(17), 3801-3805 (2000)
Effect of cyclodextrins on the stability of bilayer: A novel electrophoretic approach
Zandkarimi M, et al.
Journal of Liquid Chromatography and Related Technologies, 37(17), 2433-2443 (2014)
Preparation and application of a new glucose sensor based on iodide ion selective electrode
K Sukru, et al.
Talanta, 65(1), 87-91 (2005)
A B Packard et al.
Nuclear medicine and biology, 22(7), 887-891 (1995-10-01)
Iridium-191m is an attractive radionuclide for first-pass radionuclide angiography, but development of an 191Os/191mIr generator with high 191mIr yield has proven difficult. The use of trans-dioxobisoxalatoosmate(VI) as a parent species results in a higher initial yield (20 to 25%/mL), but
Youngjea Kang et al.
Analytical chemistry, 83(10), 3957-3962 (2011-04-20)
Contamination of heparin with oversulfated chondroitin sulfate (OSCS) became a matter of grave concern in the medical field after many fatal responses to OSCS tainted heparin products occurred during the 2007-2008 period. Even though standard lab-based analytical techniques such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service