Skip to Content
Merck
All Photos(2)

Documents

234451

Sigma-Aldrich

1-Bromohexadecane

97%

Synonym(s):

Cetyl bromide, Hexadecyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15Br
CAS Number:
Molecular Weight:
305.34
Beilstein:
773989
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

10.6 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

97%

refractive index

n20/D 1.461 (lit.)

bp

190 °C/11 mmHg (lit.)

mp

16-18 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCBr

InChI

1S/C16H33Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-16H2,1H3

InChI key

HNTGIJLWHDPAFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Bromohexadecane has been used:
  • in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains
  • as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction (MS-USAEME) method
  • in the preparation of [2-(methacryloyloxy)ethyl]dimethylhexadecylammonium bromide monomer, required for the synthesis of novel methacrylate based adsorbents
  • in the synthesis of surfactant, N-hexadecyl ethylenediamine triacetic acid (HED3A)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

350.6 °F - closed cup

Flash Point(C)

177 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Håkansson et al.
Journal of colloid and interface science, 374(1), 25-33 (2012-03-01)
The Oil Transfer Technique (OTT) was developed by Taisne et al. [1] to measure coalescence during emulsification and has been applied since in several studies. One of the main drawbacks of this technique is that it only gives a qualitative
Guomin Lin et al.
Molecules (Basel, Switzerland), 25(22) (2020-11-20)
Titanate-based bonding agents are a class of efficient bonding agents for improving the mechanical properties of composite solid propellants, a kind of special composite material. However, high solid contents often deteriorate the rheological properties of propellant slurry, which limits the
Paul D Siegel et al.
Toxicological sciences : an official journal of the Society of Toxicology, 107(1), 78-84 (2008-10-22)
The murine local lymph node assay (LLNA) is an internationally accepted assay for identification of contact allergens. The LLNA has also been used in research studies to evaluate contact allergen potency, as well as chemical structural-allergenic activity relationships. The 1-bromoalkanes
Siti Norain Harun et al.
Pharmaceutics, 13(2) (2021-01-28)
The ruthenium polypyridyl complex [Ru(dppz)2PIP]2+ (dppz: dipyridophenazine, PIP: (2-(phenyl)-imidazo[4,5-f ][1,10]phenanthroline), or Ru-PIP, is a potential anticancer drug that acts by inhibiting DNA replication. Due to the poor dissolution of Ru-PIP in aqueous media, a drug delivery agent would be a
Ning Zhang et al.
International journal of oral science, 10(2), 18-18 (2018-06-22)
Biofilms at the tooth-restoration bonded interface can produce acids and cause recurrent caries. Recurrent caries is a primary reason for restoration failures. The objectives of this study were to synthesize a novel bioactive dental bonding agent containing dimethylaminohexadecyl methacrylate (DMAHDM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service