Skip to Content
Merck
All Photos(1)

Key Documents

146552

Sigma-Aldrich

Benzyltriethylammonium chloride

99%

Synonym(s):

BTEAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(Cl)(C2H5)3
CAS Number:
Molecular Weight:
227.77
Beilstein:
3574984
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

mp

190-192 °C (dec.) (lit.)

SMILES string

[Cl-].CC[N+](CC)(CC)Cc1ccccc1

InChI

1S/C13H22N.ClH/c1-4-14(5-2,6-3)12-13-10-8-7-9-11-13;/h7-11H,4-6,12H2,1-3H3;1H/q+1;/p-1

InChI key

HTZCNXWZYVXIMZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Benzyltriethylammonium chloride is a lipophilic phase-transfer catalyst that can be used in phase-transfer catalysis (PTC) to catalyze polycondensation reactions to form high molecular weight polymers under bi-phasic conditions.[1][2][3][4]

It can also be used:
  • To activate hydroxyapatite and natural phosphate for use as a solid support for Knoevenagel condensation and Claisen-Schmidt condensation, respectively at room temperature and in the absence of a solvent.[5][6]
  • To increase the efficiency of mCPBA oxidation of sulfonimine generated from arenesulfonamide and diethyl acetal of an aromatic aldehyde to form 2-sulfonyloxaziridines.[7]
  • In combination with antimony(V) chloride to form a catalytic system for the Friedel-Crafts acylation reactions of arenes with acyl and sulfonyl chlorides.[8]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>527.0 °F - closed cup

Flash Point(C)

> 275 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase-Transfer Catalyzed Polycondensation of α,α-Dichloro-p-xylene with 2, 2-Bis (4-hydroxyphenyl) propane
Yamazaki N and Imai Y
Polymer Journal, 15(8), 603-603 (1983)
Antimony (V) Chloride-Benzyltriethylammonium Chloride Complex as an Efficient Catalyst for Friedel- Crafts Acylation Reactions
Huang A-P, et al.
Advanced Synthesis & Catalysis, 346(6), 599-602 (2004)
Antimony (V) Chloride-Benzyltriethylammonium Chloride Complex as an Efficient Catalyst for Friedel- Crafts Acylation Reactions
Huang A-P, et al.
advanced synthesis and catalysis, 346(6), 599-602 (2004)
Preparation and properties of fluorine-containing polyarylates from tetrafluoroisophthaloyl chloride and bisphenols
Kakimoto M-A, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 25(10), 2747-2753 (1987)
Hydroxyapatite as a new solid support for the Knoevenagel reaction in heterogeneous media without solvent
Sebti S, et al.
Applied Catalysis A: General, 228(1-2), 155-159 (2002)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service