Skip to Content
Merck
All Photos(2)

Key Documents

232459

Sigma-Aldrich

1,6-Diaminohexane-N,N,N′,N′-tetraacetic acid

97%

Synonym(s):

Hexamethylenedinitrilotetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO2CCH2)2N(CH2)6N(CH2CO2H)2
CAS Number:
Molecular Weight:
348.35
Beilstein:
1716834
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

232-236 °C (dec.) (lit.)

SMILES string

OC(=O)CN(CCCCCCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C14H24N2O8/c17-11(18)7-15(8-12(19)20)5-3-1-2-4-6-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

YGDVXSDNEFDTGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Behari et al.
Clinical toxicology, 16(1), 33-40 (1980-03-01)
Some polyaminocarboxylic acids were examined for their ability to mobilize chromium from certain vital organs, their subcellular fractions, and blood cells of potassium chromate administered rats. Hexamethylene 1,6-diamino tetraacetic acid (TDTA), triethylene tetramine hexaacetic acid (TTHA), and ethylene diamine di
B S Launikonis et al.
The Journal of physiology, 526 Pt 2, 299-312 (2000-07-15)
1. The role of myoplasmic [Mg2+] on Ca2+ release from the sarcoplasmic reticulum (SR) was examined in the two major types of crustacean muscle fibres, the tonic, long sarcomere fibres and the phasic, short sarcomere fibres of the fresh water
G D Lamb et al.
The Journal of physiology, 464, 629-648 (1993-05-01)
1. The effects of increased osmolality and ionic strength on the mechanism of Ca2+ release were examined in mechanically skinned skeletal muscle fibres of the toad at 23 degrees C. Ca2+ release was induced by depolarizing the transverse tubular (T-)
J M West et al.
Journal of muscle research and cell motility, 20(3), 249-264 (1999-09-03)
At early stages of muscle development, skeletal muscles contract and relax slowly, regardless of whether they are destined to become fast- or slow-twitch. In this study, we have characterised the activation profiles of developing fast- and slow-twitch muscles from a
P Schiereck et al.
Journal of muscle research and cell motility, 13(5), 573-580 (1992-10-01)
Single skinned skeletal muscle fibres were immersed in solutions containing two different levels of activator calcium (pCa: 4.4; 6.0). Sarcomere length was varied from 1.6 to 3.5 microns and recorded by laser diffraction. Slack length was 2.0 microns. Small-angle equatorial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service