Skip to Content
Merck
All Photos(1)

Key Documents

232130

Sigma-Aldrich

[Bis(trifluoroacetoxy)iodo]benzene

97%

Synonym(s):

BTI, Iodobenzene I,I-bis(trifluoroacetate), PIFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO2)2IC6H5
CAS Number:
Molecular Weight:
430.04
Beilstein:
764767
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: oxidant

mp

121-125 °C (lit.)

functional group

fluoro
iodo

SMILES string

FC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)c1ccccc1

InChI

1S/C10H5F6IO4/c11-9(12,13)7(18)20-17(6-4-2-1-3-5-6)21-8(19)10(14,15)16/h1-5H

InChI key

PEZNEXFPRSOYPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent for:
  • Pummerer-like reactions
  • Chemoselective deprotection of dimethoxybenzyl ethers
  • Mediating oxidative cycloisomerization
  • Tosyloxylation of anilides
Used for the direct α-hydroxylation of ketones under acidic conditions. Also employed as a p-fluorination reagent of 4-alkylphenols in the presence of HF·pyridine (cat. no. 184225) for synthesis of 4-fluorocyclohexa-2,5-dienones.
Reagent for the oxidation of N-acylhydrazones to 1,3,4-oxadiazoles. Promotes the cyclization of styryl amines to N-alkyl or N-aryl indoles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 35, 2541-2541 (1994)
Tetrahedron Letters, 33, 6065-6065 (1992)
Yunfei Du et al.
Organic letters, 8(26), 5919-5922 (2006-12-15)
[Structure: see text] A variety of N-arylated and N-alkylated indole derivatives were synthesized by way of a phenyliodine bis(trifluoroacetate) (PIFA)-mediated intramolecular cyclization. This novel method allows for the construction of an indole skeleton by joining the N-atom on the side
Determination of asparagine and glutamine in polypeptides using bis(I,I-trifluoroacetoxy)iodobenzene.
L M Soby et al.
Analytical biochemistry, 113(1), 149-153 (1981-05-01)
Synthetic Communications, 36, 2927-2927 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service