Skip to Content
Merck
All Photos(1)

Key Documents

103136

Sigma-Aldrich

Iodopentafluorobenzene

99%

Synonym(s):

2,3,4,5,6-Pentafluoroiodobenzene, Pentafluoroiodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6F5I
CAS Number:
Molecular Weight:
293.96
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.496 (lit.)

bp

161 °C (lit.)

density

2.204 g/mL at 25 °C (lit.)

SMILES string

Fc1c(F)c(F)c(I)c(F)c1F

InChI

1S/C6F5I/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

OPYHNLNYCRZOGY-UHFFFAOYSA-N

General description

Iodopentafluorobenzene forms supramolecular complexes with aromatic electron donors by forming halogen bonds to form discrete heterodimeric aggregates.

Application

  • Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution.
  • It was used as solvent in a study to determine singlet oxgen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery.
  • It has potential applications in plasma processing industry and in preparation of catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation and redox properties of radical ions of iodopentafluorobenzene in aqueous solution: a pulse radiolysis study
Mohan H and Mittal JP
The Journal of Physical Chemistry, 99(33), 12559-12564 (1995)
Co-crystals of iodopentafluorobenzene with nitrogen donors: 2-D molecular assemblies through halogen bonding and aryl-perfluoroaryl interactions.
Wasilewska A, et al.
CrystEngComm, 9(3), 203-206 (2007)
Chenxin Ran et al.
Chemical Society reviews, 47(12), 4581-4610 (2018-04-24)
The rapid development of solar cells (SCs) based on organic-inorganic hybrid metal triiodide perovskite (MTP) materials holds great promise for next-generation photovoltaic devices. The demonstrated power conversion efficiency of the SCs based on MTP (PSCs for short) has reached over
Influence of heavy atoms on the deactivation of singlet oxygen (1. DELTA. g) in solution
Schmidt R
Journal of the American Chemical Society, 111(18), 6983-6987 (1989)
Iodopentafluorobenzene: Electronic state spectroscopy by high-resolution vacuum ultraviolet photoabsorption and photoelectron spectroscopy.
Eden S, et al.
Chemical Physics, 359(1), 111-117 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service