Merck
  • Consequences of correlated solvation on the structures and reactivities of RLi-diamine complexes: 1,2-addition and alpha-lithiation reactions of imines by TMEDA-solvated n-butyllithium and phenyllithium.
Consequences of correlated solvation on the structures and reactivities of RLi-diamine complexes: 1,2-addition and alpha-lithiation reactions of imines by TMEDA-solvated n-butyllithium and phenyllithium.
Journal of the American Chemical Society (2002-01-10)
Jennifer L Rutherford, Daniele Hoffmann, David B Collum
ABSTRACT
6Li and (13)C NMR spectroscopic studies were carried out on [(6)Li]n-BuLi and [(6)Li]PhLi (RLi) in toluene-d(8) containing the following diamines: N,N,N',N'-tetramethylethylenediamine (TMEDA), N,N,N',N'-tetraethylethylenediamine, 1,2-dipyrrolidinoethane, 1,2-dipiperidinoethane, N,N,N',N'-tetramethylpropanediamine, trans-(R,R)-N,N,N',N'-tetramethylcyclohexanediamine, and (-)-sparteine. Dimers of general structure (RLi)(2)S(2) (S = chelating diamine) are formed in each case. Treatment of RLi with two different diamines (S and S') affords homosolvates (RLi)(2)S(2) and (RLi)(2)S'(2) along with a heterosolvate (RLi)(2)SS'. Relative binding constants and associated free energies for the sequential solvent substitutions are obtained by competing pairs of diamines. The high relative stabilities of certain heterosolvates indicate that solvent binding to the RLi dimer can be highly correlated. Rate studies of both the 1,2-addition of RLi/TMEDA to the N-isopropylimine of cyclohexane carboxaldehyde and the RLi/TMEDA-mediated alpha-lithiation of the N-isopropylimine of cyclohexanone reveal monomer-based transition structures, [(RLi)(TMEDA)(imine)], in all cases. The complex relationships of solvent binding constants and relative reactivities toward 1,2-additions and alpha-lithiations are discussed.
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane