Skip to Content
Merck
All Photos(2)

Key Documents

230030

Sigma-Aldrich

Cesium sulfate

99.99% trace metals basis

Synonym(s):

Caesium sulphate, Dicesium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs2O4S
CAS Number:
Molecular Weight:
361.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

impurities

≤200.0 ppm Trace Metal Analysis

mp

1019 °C (lit.)

density

4.243 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[Cs+].[O-]S([O-])(=O)=O

InChI

1S/2Cs.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

FLJPGEWQYJVDPF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Gruenwedel et al.
Biopolymers, 7(4), 557-570 (1969-01-01)
When DNA's of differing GC:AT base ratios, e.g. synthetic poly dAT, T4 DNA,calf thymus DNA, E. coli DNA, and M. lysodeikticus DNA, are heat-denatured at neutral pH in increasing concentrations of N(a)(2)SO(4) or C(s)(2)SO(4) as supporting electrolytes,the variation of melting
F A Aly et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 17(11), 1257-1261 (2002-01-05)
A flow-injection (FI) methodology using tris(2,2'-dipyridyl)ruthenium(II), [Ru(dipy)3(2+)], chemiluminescence (CL) was developed for the rapid and sensitive determination of three thioxanthene derivatives, namely zuclopenthixol hydrochloride, flupentixol hydrochloride and thiothixene. The method is based on the CL reaction of the studied thioxanthenes
F M Ashcroft et al.
The Journal of physiology, 359, 269-291 (1985-02-01)
The non-penetrating potentiometric dyes NK2367 and WW375 were used to investigate the effect of inward rectification on the weighted-average tubular membrane potential in single frog muscle fibres voltage clamped using a three-Vaseline-gap method. In 100 mM-K solution, when inward rectification
A Miyakawa et al.
Neuroscience, 134(2), 595-604 (2005-06-25)
Effects of neuropeptide Y (NPY) on substantia gelatinosa neurons were investigated in adult rat spinal cord slices using blind whole-cell patch-clamp technique. Bath application of NPY (1 microM) induced a membrane hyperpolarization, resulting in a suppression of the dorsal root
A K Attri et al.
Analytical biochemistry, 159(1), 88-95 (1986-11-15)
We present a technique for analytical buoyant density sedimentation of nucleic acids which is performed in a preparative ultracentrifuge, in contrast to an analytical ultracentrifuge. Following centrifugation in a preparative rotor, small cylindrical quartz tubes are optically scanned; upon completion

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service