Skip to Content
Merck
All Photos(2)

Key Documents

225460

Sigma-Aldrich

Diironnonacarbonyl

98%

Synonym(s):

Enneacarbonyldiiron, Iron enneacarbonyl, Nonacarbonyldiiron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(CO)9
CAS Number:
Molecular Weight:
363.78
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

core: iron
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=C1[Fe]2C(=O)[Fe]1C2=O

InChI

1S/9CO.2Fe/c9*1-2;;

InChI key

JCXLZXJCZPKTBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reacts with phenylsilanes to give triply bridged diiron complexes.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Sol. 2 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerber, R. C. et al.R. C.; Ribakove, E. C.
Organometallic News, 10, 2848-2848 (1991)
Gree, R.
Synthesis, 341-341 (1989)
Luca Bertini et al.
The journal of physical chemistry. A, 111(48), 12152-12162 (2007-11-09)
The singlet and triplet state potential energy surfaces (PES) of Fe2(CO)8 are explored by means of density functional theory (DFT) techniques. The two PES have different global mimima: the dibriged C(2v) isomer for the singlet and the unbridged D(2d) isomer
Nu Xiao et al.
Dalton transactions (Cambridge, England : 2003), (4)(4), 603-608 (2006-01-13)
[{Micro-(phthalazine-N2:N3)}Fe2(micro-CO)(CO)6](1) reacts with organolithium reagents, RLi (R = CH3, C6H5, p-CH3C6H4, p-CH3OC6H4, p-CF3C6H4, p-C6H5C6H4), followed by treatment with Me3SiCl to give the novel diiron carbonyl complexes with a saturated N-N six-membered diazane ring ligand, [{C6H4CH(R)NNCH2}Fe2(C=O)(CO)6](2, R = CH3; 3, R
Simons, R.S. Tessier, C.A.
Organometallics, 15, 2604-2604 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service