Skip to Content
Merck
All Photos(2)

Documents

218359

Sigma-Aldrich

(S)-(−)-Perillic acid

95%

Synonym(s):

4-Isopropenylcyclohexene-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

flakes

optical activity

[α]20/D −102°, c = 2 in methanol

mp

129-131 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)[C@H]1CCC(=CC1)C(O)=O

InChI

1S/C10H14O2/c1-7(2)8-3-5-9(6-4-8)10(11)12/h5,8H,1,3-4,6H2,2H3,(H,11,12)/t8-/m1/s1

InChI key

CDSMSBUVCWHORP-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-Perillic acid is a perillyl derivative with potent antimicrobial and anticancer activity. It can be prepared via bio-oxidation of R-(+)-limonene.

Application

(S)-(−)-Perillic acid (PA) can be used as a starting material for the preparation of:,·        
  • Aryl amides, 4-(prop-1-en-2-yl)-N-(3-(trifluoromethyl)phenyl)cyclohex-1-ene-1-carboxamide and N-(4-(4-amino-2-methylphenethyl)-3-methylphenyl)-4-(prop-1-en-2-yl)cyclohex-1-ene-1-carboxamide, as in vitro antiproliferative active agents.
  •  Machaeriols and cannabinoid-related compounds as antimalarial agents.   
  • Tricyclic-β-lactone by treating with 1-chloro-N,N-2-trimethylpropenylamine and dimethylketomalonate, followed by addition of 9-azajulolidine.

Biochem/physiol Actions

Interferes with activity of p21ras and other small G proteins by inhibiting post-translational cysteine isoprenylation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel N-arylamide derivatives of (S)-perillic acid ((S)-PA): in vitro and in vivo cytotoxicity and antitumor evaluation
Mukhtar YM, et al.
Royal Society of Chemistry Advances, 9(35), 19973-19982 (2019)
Natural product derivatization with β -lactones, β -lactams and epoxides toward `infinite?binders
Jouanneau M, et al.
Tetrahedron, 75(24), 3348-3354 (2019)
Bioconversion of R-(+)-limonene to perillic acid by the yeast Yarrowia lipolytica.
Ferrara MA, et al.
Brazilian Journal of Microbiology, 44(4), 1075-1080 (2013)
Felix Klotter et al.
Angewandte Chemie (International ed. in English), 54(29), 8547-8550 (2015-06-17)
Short and highly efficient stereoselective syntheses provide machaeriols and cannabinoids in a divergent approach starting from a common precursor, commercially available (S)-perillic acid. Key features of the novel strategy are a stereospecific palladium-catalyzed decarboxylative arylation and a one-pot sequence comprising

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service