Skip to Content
Merck
All Photos(2)

Key Documents

214973

Sigma-Aldrich

Diisobutylaluminum hydride solution

1.0 M in methylene chloride

Synonym(s):

DIBAL, DIBAL-H

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein:
4123663
MDL number:
UNSPSC Code:
12352001
eCl@ss:
38120609
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in methylene chloride

density

1.23 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)C[AlH]CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;

InChI key

AZWXAPCAJCYGIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisobutylaluminum hydride is a reducing agent for many functional groups commonly used in organic synthesis.

Application

Diisobutylaluminum hydride solution (1 M in dichloromethane) has been used in the conversion of esters into α-fluoro-α,β-unsaturated esters in the presence of fluorocarboalkoxy-substituted dialkyl phosphonate anions via Horner-Wadsworth-Emmons reaction. It can be used in the multi-step synthesis of (S)-2-methyl tetrahydropyridine-N-oxide, a key structural moiety of bio-active compounds like himbacine and solenopsin-A.
Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction-olefination of esters: a new and efficient synthesis of. alpha.-fluoro. alpha.,. beta.-unsaturated esters.
Thenappan A
The Journal of Organic Chemistry, 55(15), 4639-4642 (1990)
Diisobutylaluminum Hydride.
Galatsis P
e-EROS Encyclopedia of Reagents for Organic Synthesis (2008)
An enantioselective route to trans-2, 6-disubstituted piperidines
Chackalamannil S
Tetrahedron, 53(32), 11203-11210 (1997)
Reaction of diisobutylaluminum hydride with selected organic compounds containing representative functional groups
Yoon N M
The Journal of Organic Chemistry, 50(14), 2443-2450 (1985)
D J Kopecky et al.
The Journal of organic chemistry, 65(1), 191-198 (2000-05-18)
An optimized protocol for the DIBALH reductive acetylation of acyclic esters and diesters is described. This reductive acetylation procedure allows a wide variety of esters to be converted into the corresponding alpha-acetoxy ethers in good to excellent yields. It was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service