Skip to Content
Merck
All Photos(2)

Key Documents

214914

Sigma-Aldrich

N,N-Dimethylmethyleneiminium iodide

98%

Synonym(s):

N,N-Dimethylmethyleneammonium iodide, Eschenmoser’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=N+(CH3)2I-
CAS Number:
Molecular Weight:
185.01
Beilstein:
1731022
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

219 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear to slightly hazy, colorless to yellow

functional group

amine

SMILES string

[I-].C[N+](C)=C

InChI

1S/C3H8N.HI/c1-4(2)3;/h1H2,2-3H3;1H/q+1;/p-1

InChI key

VVDUZZGYBOWDSQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylmethyleneiminium iodide is useful reagent for many synthetic applications, especially aminomethylation and conversion of ketones to α,β−unsaturated enones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayuki Inoue et al.
The Journal of organic chemistry, 72(8), 3065-3075 (2007-03-16)
(-)-Merrilactone A [(-)-1], isolated from Illicium merrillianum in 2000, possesses neurite outgrowth activity in cultures of fetal rat cortical neurons, and, therefore, is expected to show therapeutic potential for the treatment of neurodegeneration associated with Alzheimer's and Parkinson's diseases. Apart
A new method using 2-chloro-4, 6-dimethoxy-1, 3, 5-triazine for facile elimination of dimethylamino group in Eschenmoser's methylenation for synthesis of a, ?-unsaturated esters.
Yamada K, et al.
Tetrahedron Letters, 54(13), 1753-1760 (2013)
Tetrahedron Asymmetry, 5, 921-921 (1994)
J. Prakt. Chem./Chem.-Ztg., 336, 91-91 (1994)
Journal of the Chemical Society. Perkin Transactions 1, 3295-3295 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service