Skip to Content
Merck
All Photos(1)

Key Documents

432741

Sigma-Aldrich

(S)-(−)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde

95%

Synonym(s):

(−)-N-Boc-N,O-isopropylidene-L-serinal, tert-Butyl (S)-(−)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, Garner’s aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
Beilstein:
3591324
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

optical activity

[α]20/D −90°, c = 1 in chloroform

refractive index

n20/D 1.445 (lit.)

bp

83-88 °C/1 mmHg (lit.)

density

1.06 g/mL at 25 °C (lit.)

functional group

aldehyde
ether

SMILES string

[H]C(=O)[C@@H]1COC(C)(C)N1C(=O)OC(C)(C)C

InChI

1S/C11H19NO4/c1-10(2,3)16-9(14)12-8(6-13)7-15-11(12,4)5/h6,8H,7H2,1-5H3/t8-/m1/s1

InChI key

PNJXYVJNOCLJLJ-MRVPVSSYSA-N

Application

Widely utilized in the asymmetric synthesis of natural products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoemann, M.Z. et al.
Tetrahedron Letters, 37, 953-953 (1996)
Guibourdenche, C. et al.
Tetrahedron Asymmetry, 4, 2041-2041 (1993)
Roush, W.R. Hunt, J.A.
The Journal of Organic Chemistry, 60, 798-798 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service