All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4ClNO · HCl
CAS Number:
Molecular Weight:
178.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
151-157 °C (lit.)
functional group
acyl chloride
SMILES string
Cl[H].ClC(=O)c1cccnc1
InChI
1S/C6H4ClNO.ClH/c7-6(9)5-2-1-3-8-4-5;/h1-4H;1H
InChI key
MSYBLBLAMDYKKZ-UHFFFAOYSA-N
Related Categories
Application
Nicotinoyl chloride hydrochloride was used in the preparation of :
- 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil
- 7,16-dinicotinoylated and/or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes
- bis(ethylenedithio)tetrathiafulvalene derivatives
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of bis(ethylenedithio) tetrathiafulvalene derivatives with metal ion ligating centres.
Griffiths J-P, et al.
Tetrahedron Letters, 44(15), 31327-31331 (2003)
Shuang Li et al.
Journal of the mechanical behavior of biomedical materials, 102, 103521-103521 (2019-12-27)
In this research, a bio-based monomer 1,3-bis(methacryloyloxy)propyl-carbonyl- hexylpyridinium bromide (QANMA) that derived from niacin was synthesized and incorporated into Bisphenol A glycidyl methacrylate (Bis-GMA)/triethylene glycol dimethacrylate (TEGDMA) (50 wt/50 wt) with a series of mass fraction as antibacterial agent. The double bond
K W Morin et al.
Carbohydrate research, 249(1), 109-116 (1993-10-18)
1-(2-Deoxy-2-fluoro-beta-D-ribofuranosyl-(E)-5-(2-iodovinyl)uracil (IVFRU) was coupled to a dihydropyridine <---> pyridinium salt redox chemical-delivery system (CDS) via a cleavable sugar-ester linkage as a site-directed approach to increase diffusion of the parent nucleoside into the central nervous system. Treatment of 1-(2-deoxy-2-fluoro-beta-D-ribofuranosyl)uracil with Bu(t)Me(2)SiCl
Synthesis and characterization of 7, 16-dinicotinoyl-and 7, 16-diisonico-tinoyltetraaza [14] annulene and their nickel (II) and copper (II) complexes.
Sakata K and Itoh M.
Journal of Heterocyclic Chemistry, 29(4), 921-926 (1992)
Wenqiang Tan et al.
Carbohydrate polymers, 157, 236-243 (2016-12-19)
Based on cuprous-catalyzed azide-alkyne cycloaddition (CuAAC), starch derivative bearing 1,2,3-triazole and pyridine (II) was prepared and subsequently followed by alkylation with iodomethane to synthesize starch derivative bearing 1,2,3-triazolium and pyridinium (III). The antifungal activities of starch derivatives against Colletotrichum lagenarium
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service