Skip to Content
Merck
All Photos(1)

Key Documents

210145

Sigma-Aldrich

2,2-Dimethyl-1,3-dioxane-4,6-dione

2,2-Dimethyl-1,3-dioxane-4,6-dione

98%

Synonym(s):

Malonic acid cyclic isopropylidene ester, Meldrum’s acid, cycl-Isopropylidene malonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
Beilstein:
117310
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

92-96 °C (lit.)

solubility

dioxane: soluble 5%, clear to very slightly hazy, colorless to faintly yellow

functional group

ester
ketal

storage temp.

2-8°C

SMILES string

CC1(C)OC(=O)CC(=O)O1

InChI

1S/C6H8O4/c1-6(2)9-4(7)3-5(8)10-6/h3H2,1-2H3

InChI key

GXHFUVWIGNLZSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethyl-1,3-dioxane-4,6-dione (Meldrum′s acid) is widely used in organic synthesis, especially for multiple C-C bond formations due to its adequate acidity (pKa 4.83) and steric rigidity[1]. Knoevenagel condensation reaction between aldehydes and Meldrum′s acid are accelerated in ionic liquids.[2]

Meldrum′s acid is used as a valuable starting material to synthesize heterocycles and as intermediates in organic synthesis reactions.[3][4]

Application

2,2-Dimethyl-1,3-dioxane-4,6-dione was used in the synthesis of:
  • macrocyclic β-keto lactone[5]
  • 4-pyridyl-substituted heterocycles[6]
  • 2-substituted indoles[7]
  • isofraxidin.[8]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New multicomponent domino reactions (MDRs) in water: highly chemo-, regio-and stereoselective synthesis of spiro {[1, 3] dioxanopyridine}-4, 6-diones and pyrazolo [3, 4-b] pyridines.
Ma N, et al.
Green Chemistry, 12?(8), 1357-1361 (2010)
Songlei Zhu et al.
Molecules (Basel, Switzerland), 17(12), 13856-13863 (2012-11-24)
A series of 4-aryl-6-methyl-3,4-dihydro-2H-pyrano[3,2-c]quinolin-2,5(6H)-diones were synthesized via the three-component reactions of aromatic aldehydes, 4-hydroxy-1-methylquinolin-2(1H)-one, and Meldrum's acid catalyzed by L-proline. The structures of the products were identified by spectroscopic analysis. A mechanism for this three-component reaction catalyzed by L-proline was
Davood Nematollahi et al.
Chemical & pharmaceutical bulletin, 58(1), 23-26 (2010-01-05)
Electrochemical oxidation of catechols in the presence of phenyl-Meldrum's acid as a nucleophile in aqueous solution has been studied in detail by means of cyclic voltammetry and controlled potential coulometry. The results indicate that the o-benzoquinone derived from catechols participates
The synthesis of β-keto lactones via cyclization of β-keto ester dianions or the cyclization of Meldrum's acid derivatives.
Lermer L, et al.
Canadian Journal of Chemistry, 70(5), 1427-1445 (1992)
Characterization of Meldrum?s acid derivative 5-(5-Ethyl-1, 3, 4-thiadiazol-2-ylamino) methylene-2, 2-dimethyl-1, 3-dioxane-4, 6-dione by Raman and FT-IR spectroscopy and DFT calculations
De Toledo TA, et al.
Journal of Molecular Structure, 37-42, 1091-1091 (2015)

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service