Skip to Content
Merck
All Photos(3)

Key Documents

204749

Sigma-Aldrich

Ammonium hexafluorotitanate

99.99% trace metals basis

Synonym(s):

Ammonium fluotitanate, Ammonium titanium fluoride, Diammonium hexafluorotitaniumdiuide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2TiF6
CAS Number:
Molecular Weight:
197.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: titanium

impurities

≤150.0 ppm Trace Metal Analysis

solubility

H2O: slightly soluble(lit.)

SMILES string

[H][N+]([H])([H])[H].[H][N+]([H])([H])[H].F[Ti--](F)(F)(F)(F)F

InChI

1S/6FH.2H3N.Ti/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

NMGYKLMMQCTUGI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenichiro Hayashi et al.
Materials (Basel, Switzerland), 13(1) (2019-12-28)
Titania (TiO2) has attracted much attention recently for reducing bacterial diseases by the generation of reactive oxygen species (ROS) under UV irradiation. However, demand for higher photocatalytic activity due to higher recombination of electron and hole remains. The aims of
Daejong Yang et al.
Langmuir : the ACS journal of surfaces and colloids, 34(14), 4132-4141 (2018-03-16)
We report ZnO nanowire- and TiO2 nanotube-based light sensors on flexible polymer substrates fabricated by localized hydrothermal synthesis and liquid phase deposition (LPD). This method realized simple and cost-effective in situ synthesis and integration of one-dimensional ZnO and TiO2 nanomaterials.
Lei Zhou et al.
Journal of the American Chemical Society, 130(4), 1309-1320 (2008-01-10)
Uniform mesocrystals of TiO2 (anatase) have been prepared from mesocrystals of NH4TiOF3. NH4TiOF3 was synthesized from an aqueous solution containing (NH4)2TiF6 and H3BO3 in the presence of a nonionic surfactant Brij 56, Brij 58, or Brij 700, at low temperatures.
Jin Shen et al.
Nature communications, 11(1), 6259-6259 (2020-12-09)
Three-dimensional metal-organic frameworks (MOFs) are cutting-edge materials in the adsorptive removal of trace gases due to the availability of abundant pores with specific chemistry. However, the development of ideal adsorbents combining high adsorption capacity with high selectivity and stability remains
[Effect of titanium fluoride compounds on human enamel].
S Iino
Koku Eisei Gakkai zasshi, 31(5), 63-78 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service