Skip to Content
Merck
All Photos(2)

Key Documents

238007

Sigma-Aldrich

Potassium hexafluoroantimonate(V)

99%

Synonym(s):

Potassium antimony fluoride, Potassium hexafluorostibanuide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSbF6
CAS Number:
Molecular Weight:
274.85
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: antimony

bp

1505 °C/1 atm (lit.)

mp

846 °C (lit.)

SMILES string

[K+].F[Sb-](F)(F)(F)(F)F

InChI

1S/6FH.K.Sb/h6*1H;;/q;;;;;;+1;+5/p-6

InChI key

LQKKGPUEJBHVHZ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Merve Kara et al.
Macromolecular rapid communications, 36(23), 2070-2075 (2015-09-12)
A new phenacyl-type photoinitiator based on ethyl carbazole as a long wavelength photo-initiator is developed for free radical polymerization. Phenacyl ethyl carbazolium hexafluoroantimonate (PECH) photoinitiator is synthesized in a two-step, one-pot manner by quaternizing ethyl carbazole with phenacyl bromide and

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service