Skip to Content
Merck
All Photos(4)

Key Documents

202827

Sigma-Aldrich

Bismuth(III) oxide

powder, 99.999% trace metals basis

Synonym(s):

Dibismuth trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi2O3
CAS Number:
Molecular Weight:
465.96
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38080302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

SMILES string

O=[Bi]O[Bi]=O

InChI

1S/2Bi.3O

InChI key

WMWLMWRWZQELOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Mass spectrometry standards: A significant advancement was made in developing an enantioselective high-performance liquid chromatography-tandem mass spectrometry method for the quantitative determination of methorphan and its O-demethylated metabolite, dextrorphan-d₃, in human blood, specifically for forensic applications in post-mortem samples. This method underscores the utility of stable isotope-labeled compounds like dextrorphan-d₃ in precise pharmacological research (Lo Faro et al., 2023).

Features and Benefits

Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sanghi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 94-99 (2011-09-06)
Glasses with composition 20CdO·xBi(2)O(3)·(79.5-x)B(2)O(3) (15≤x≤35, x in mol%) containing 0.5 mol% of Er(3+) ions were prepared by melt-quench technique (1150°C in air). The amorphous nature of the glasses was confirmed by X-ray diffraction. The spectroscopic properties of the glasses were
Atsushi Satsuma et al.
Sensors (Basel, Switzerland), 11(2), 2155-2165 (2012-02-10)
Mixed potential sensors were fabriated using yttria-stabilized zirconia (YSZ) as a solid electrolyte and a mixture of Au and various metal oxides as a sensing electrode. The effects of calcination temperature ranging from 600 to 1,000 °C and acid-base properties
Lu You et al.
ACS nano, 6(6), 5388-5394 (2012-05-18)
The novel strain-driven morphotropic phase boundary (MPB) in highly strained BiFeO(3) thin films is characterized by well-ordered mixed phase nanodomains (MPNs). Through scanning probe microscopy and synchrotron X-ray diffraction, eight structural variants of the MPNs are identified. Detailed polarization configurations
V S Kopp et al.
Acta crystallographica. Section A, Foundations of crystallography, 68(Pt 1), 148-155 (2011-12-22)
X-ray diffraction from films consisting of layers with different thicknesses, structures and chemical contents is analysed. The disorder is described by probabilities for different sequences of layers. Closed analytical expressions for the diffracted X-ray intensity are obtained when the layers
L M Formosa et al.
Dental materials : official publication of the Academy of Dental Materials, 28(5), 584-595 (2012-03-14)
Tricalcium silicate is the major constituent phase in mineral trioxide aggregate (MTA). It is thus postulated that pure tricalcium silicate can replace the Portland cement component of MTA. The aim of this research was to evaluate the microstructure and surface

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service