Skip to Content
Merck
All Photos(2)

Key Documents

202746

Sigma-Aldrich

Barium fluoride

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaF2
CAS Number:
Molecular Weight:
175.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

Quality Level

Assay

99.99% trace metals basis

form

solid

impurities

≤150.0 ppm Trace Metal Analysis

bp

2260 °C (lit.)

density

4.89 g/mL at 25 °C (lit.)

SMILES string

F[Ba]F

InChI

1S/Ba.2FH/h;2*1H/q+2;;/p-2

InChI key

OYLGJCQECKOTOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Primary glass modifier in fluorozirconate glasses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jane G Zapka et al.
American journal of preventive medicine, 23(1), 28-35 (2002-07-03)
Developing effective programs to promote colorectal cancer (CRC) screening requires understanding of the effect of healthcare system factors on access to screening and adherence to guidelines. This study assessed the role of insurance status, type of plan, the frequency of
W H Wong
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 29(3), 338-347 (1988-03-01)
Bismuth germanate (BGO) and barium fluoride (BaF2) scintillators are presently used in positron emission tomegraphy (PET) cameras. This study evaluates the important PET performance parameters of image resolution, true sensitivity, scatter background, accidental background, and realistic maximum radioactivity in the
T J Bastow et al.
Solid state nuclear magnetic resonance, 6(1), 95-100 (1996-02-01)
The X-ray storage phosphor materials BaFC1 and BaFBr differ greatly in their 137Ba nuclear electric quadrupole interaction, despite a common crystal structure. The quadrupole coupling constant in BaFCl is relatively small and increases rapidly with temperature, in an unusual manner:
Wendy Patterson et al.
Optics express, 16(3), 1704-1710 (2008-06-11)
We report laser-induced cooling with thulium-doped BaY2F8 single crystals grown using the Czochralski technique. The spectroscopic characterization of the crystals has been used to evaluate the laser cooling performance of the samples. Cooling by 3 degrees below ambient temperature is
Allosteric binding of halide anions by a new dimeric interpenetrated coordination cage.
Sabrina Freye et al.
Angewandte Chemie (International ed. in English), 51(9), 2191-2194 (2012-01-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service