Skip to Content
Merck
All Photos(1)

Key Documents

202339

Sigma-Aldrich

Poly(propylene glycol)

average Mn ~2,000

Synonym(s):

PPG, Poly(propylene oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OCH(CH3)CH2]nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
vapor pressure:
<0.01 mmHg ( 20 °C)

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

mol wt

average Mn ~2,000

contains

130-190 ppm proprietary phenolic antioxidant

refractive index

n20/D 1.451

viscosity

300 cSt(25 °C)(lit.)

hydroxyl value

56 mg KOH/g

density

1.005 g/mL at 25 °C

SMILES string

CC(O)CO

InChI

1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3

InChI key

DUFKCOQISQKSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(propylene glycol) (PPG) is a polyether. It is prepared by ring-opening polymerization of propylene oxide. PPG is less toxic and used as a dispersant in leather finishing, as a surfactant, wetting agent, and in drug formulations.

Application

Poly(propylene glycol) can be used as a:
  • Corrosion inhibitor for mild steel in acid solution.
  • Starting material to synthesize epoxy resin/polyurethane hybrid networks via frontal polymerization.
It can also be used to prepare poly(PEG/PPG/PTMC urethane) thermogels for sustained release of doxorubicin.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

445.0 °F - closed cup

Flash Point(C)

229.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio Poggio et al.
Journal of endodontics, 38(9), 1239-1243 (2012-08-16)
The objective of this study was to evaluate and compare in vitro the decalcifying capability on root canal dentin of 4 different irrigating solutions. Twenty-five freshly extracted maxillary central incisors were selected. The canals were prepared to obtain a total
Zhiyong Zhao et al.
Chemical communications (Cambridge, England), 48(78), 9753-9755 (2012-08-25)
The designed DNA sequences can make DNA-b-PPO undergo in situ transition between diblock and triblock upon pH changes, consequently, induce a morphology-shifting from spherical micelles to nanofibers. This process is reversible and the assembled structures have been characterized by CD
Daimei Chen et al.
Journal of hazardous materials, 235-236, 186-193 (2012-08-14)
TiO(2) pillared clay materials were prepared by montmorillonite (Mt) and acidic solutions of hydrolyzed Ti alkoxides in the presence of high-molecular-weight polyoxypropylene (POP)-backboned di-quaternary salts (POP). The as-prepared materials were characterized by means of XRD, FTIR, TG-DTA, XRF, specific surface
James E Hudson et al.
Tissue engineering. Part A, 17(17-18), 2279-2289 (2011-05-06)
Tissue engineering approaches are currently being investigated for the restoration of myocardial function in heart failure patients, most commonly by combining cells with a substrate to form myocardial-like constructs (MCs). The final properties of these constructs are dependant on the
Jessalyn Cortese et al.
Journal of the American Chemical Society, 134(8), 3671-3674 (2012-02-11)
We show here that complementary interactions can suppress mesoscopic order and thus lead to a counterintuitive change in material properties. We present results for telechelic supramolecular polymers based on poly(propylene oxide) (PPO), thymine (Thy), and diaminotriazine (DAT). The self-complementary systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service