Skip to Content
Merck
All Photos(4)

Documents

201332

Sigma-Aldrich

Zincon sodium salt

Dye content ≥75 %

Synonym(s):

2-Carboxy-2′-hydroxy-5′-sulfoformazyl-benzene sodium salt, 2-[5-(2-Hydroxy-5-sulfophenyl)-3-phenyl-1-formazyl]benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H16N4NaO6S · xNa
CAS Number:
Beilstein:
728403
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, ≥75%

mp

250 °C

solubility

0.1 M NaOH: 1 mg/mL

λmax

523 nm

ε (extinction coefficient)

≥10000 at 489-495 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].OC(=O)c1ccccc1\N=N\C(=N\Nc2cc(ccc2O)S([O-])(=O)=O)c3ccccc3

General description

Zincon is a formazan dye, that serves as a metal indicator and is applicable in chelometric titration. It is used in spectrometric analysis. The metal-zincon complex formed shows a prominent absorption band in the visible region. Zincon is also used in solid support electrophoresis for staining zinc proteins.

Application

Zincon sodium salt has been used to locate zinc in plant tissues by light microscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histochemical Detection of Zn in Plant Tissues
Seregin Ilya, et al.
Bio-protocol, 5, 349-349 (2015)
Molecular Spectroscopy, 195-195 (1976)
Handbook of Biochemistry and Molecular Biology, 2706-2706 (2018)
CRC Handbook of Organic Analytical Reagents, 349-349 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service