Skip to Content
Merck
All Photos(3)

Key Documents

198080

Sigma-Aldrich

Methyl Green

zinc chloride salt, certified by the Biological Stain Commission, Dye content 85 %

Synonym(s):

Ethyl Green

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H35BrClN3 · ZnCl2
CAS Number:
Molecular Weight:
653.24
Colour Index Number:
42590
Beilstein:
3914106
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, 85%

technique(s)

titration: suitable

pH

0.2-1.8, yellow to blue

mp

>300 °C (lit.)

solubility

H2O: 1 mg/mL

λmax

423 nm (2nd)
629 nm

ε (extinction coefficient)

≥11100 at 417-425 in H2O at 0.005 g/L
≥16000 at 311-319 in H2O at 0.005 g/L
≥60000 at 626-634 in H2O at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Br-].Cl[Zn]Cl.CC[N+](C)(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](\C)C

InChI

1S/C27H35N3.BrH.3ClH.Zn/c1-8-30(6,7)26-19-13-23(14-20-26)27(21-9-15-24(16-10-21)28(2)3)22-11-17-25(18-12-22)29(4)5;;;;;/h9-20H,8H2,1-7H3;4*1H;/q+2;;;;;+2/p-4

InChI key

RRKSQWOQZFIQKW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Green is a basic dicationic dye and is composed of triphenylmethane. It is suitable for the detection of DNA and RNA. It acts as a photochromophore and used to activate gelatinous films. At a very high pH, methyl green forms an alkaline solution and gets converted to a colorless carbinol base. In the presence of hydroxide anion, methyl green produces crystal violet dye and ethanol.

Suitability

Suitable for use as a DNA stain.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic studies of the photocatalytic degradation of methyl green: an investigation of products of the decomposition processes
Chen CC and Lu CS
Environmental Science & Technology, 41(12), 4389-4396 (2007)
R.D. Lillie Ed.
Conn?s Biological Stains, 282-282 (1977)
Virchow?s Arch. (Pathol. Anat.), 157, 19-19 (1899)
The problem of a standardized technic for the methyl-green-pyronin stain.
E B TAFT
Stain technology, 26(3), 205-212 (1951-07-01)
Journal of the American Medical Association, 76, 240-240 (1921)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service