Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl Green
Synonym(s):
Ethyl Green
Linear Formula:
C
27
H
35
BrClN
3
· ZnCl
2
CAS Number:
7114-03-6
Molecular Weight:
653.24
EC Number:
230-415-4
Products
Product Category
stains (4)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
67060
Methyl Green,
zinc chloride salt, for microscopy (Bact., Bot., Hist.)
Expand
View Pricing
323829
Methyl Green,
zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Expand
View Pricing
198080
Methyl Green,
zinc chloride salt, certified by the Biological Stain Commission, Dye content 85 %
Expand
View Pricing
M8884
Methyl Green,
~85% purity, powder
Expand
View Pricing