Skip to Content
Merck
All Photos(1)

Key Documents

196746

Sigma-Aldrich

2,3,4,5,6-Pentafluorobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5CH2OH
CAS Number:
Molecular Weight:
198.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

114-115 °C/60 mmHg (lit.)

mp

37-38 °C (lit.)

SMILES string

OCc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C7H3F5O/c8-3-2(1-13)4(9)6(11)7(12)5(3)10/h13H,1H2

InChI key

PGJYYCIOYBZTPU-UHFFFAOYSA-N

General description

Structures of complex of horse liver alchohol dehydrogenase enzyme with 2,3,4,5,6-pentafluorobenzyl alcohol has been investigated by X-ray crystallography.

Application

2,3,4,5,6-Pentafluorobenzyl alcohol was used in the synthesis of pentafluorobenzyloxy substituted metal-free phthalocyanine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

V G Voinov et al.
Analytical chemistry, 76(10), 2951-2957 (2004-05-18)
A prototype gas chromatograph (GC) electron monochromator (EM) reflectron time-of-flight (TOF) mass spectrometer has been constructed and demonstrated to simultaneously record four-dimensional resonant electron capture (REC) mass spectra (m/z, ion-intensity, electron-energy, and retention time) of electron-capturing compounds in real time.
S Ramaswamy et al.
Biochemistry, 33(17), 5230-5237 (1994-05-03)
Structures of the enzyme complexed with NAD+ and 2,3,4,5,6-pentafluorobenzyl alcohol were determined by X-ray crystallography at a resolution of 2.1 A and to a refinement R value of 18.3% for a monoclinic (P2(1)) form and to 2.4 A and an
C Oman
Environmental science & technology, 35(1), 232-239 (2001-05-16)
Emissions of organic compounds from landfills depend on the fate of the compounds inside the landfills. This field study was used to investigate the fate in landfills of organic compounds having different physical, chemical, and biological characteristics. For this purpose
Jon K Rubach et al.
Biochemistry, 41(52), 15770-15779 (2002-12-27)
The relationship between substrate mobility and catalysis was studied with wild-type and Phe93Ala (F93A) horse liver alcohol dehydrogenase (ADH). Wild-type ADH binds 2,3,4,5,6-pentafluorobenzyl alcohol in one position as shown by X-ray results, and (19)F NMR shows five resonances for the
Novel phthalocyanines with pentafluorobenzyloxy-substituents.
Selcukoglu M and Hamuryudan E.
Dyes and Pigments, 74(1), 17-20 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service