Skip to Content
Merck
All Photos(1)

Documents

191752

Sigma-Aldrich

3-Phenoxybenzaldehyde

98%

Synonym(s):

3-Phenoxybenzaldehyde, 5-Phenoxybenzaldehyde, m-(Phenyloxy)benzaldehyde, m-Phenoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4CHO
CAS Number:
Molecular Weight:
198.22
Beilstein:
511662
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.595 (lit.)

bp

169-169.5 °C/11 mmHg (lit.)

density

1.147 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1cccc(Oc2ccccc2)c1

InChI

1S/C13H10O2/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-10H

InChI key

MRLGCTNJRREZHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Enantioselective autoinduction during asymmetric hydrocyanation of 3-phenoxybenzaldehyde catalyzed by cyclo[(R)-phenylalanyl-(R)-histidyl] has been investigated. It undergoes hydrogenation catalyzed by Au/Pt bimetallic core/shell nanoparticles to yield 3-phenoxyphenyl methanol.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup - (External MSDS)

Flash Point(C)

113 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Voltammetric behaviour of the synthetic pyrethroid lambda-cyhalothrin and its determination in soil and well water.
Oudou HC, et al.
Analytica Chimica Acta, 523(1), 69-74 (2004)
Jan von Langermann et al.
Bioprocess and biosystems engineering, 31(3), 155-161 (2008-01-22)
The application of unusual high pH-values within enzymatic cyanohydrin synthesis has been investigated. Usually enzymatic cyanohydrin synthesis in two-phase systems requires low pH-values within the aqueous phase to suppress the non-enzymatic side reaction. In contrast, we investigated the usage of
Sh T Iskandarova
Gigiena i sanitariia, (3)(3), 6-8 (1996-05-01)
The probable odour threshold of 3-phenoxybenzaldehyde (3-PBA) in the ambient air was estimated at 0.225 mg/m3. The maximum permissible concentration (MPC) of 3-PBA on a single exposure was recommended to be 0.09 mg/m3. The mean daily MPC of 3-PBA was
Soamrutai Boonsuepsakul et al.
Archives of insect biochemistry and physiology, 69(1), 13-21 (2008-07-11)
Metabolism by cytochrome P450 monooxygenases is a major mechanism implicated in resistance of insects to insecticides, including pyrethroids. We previously isolated the cytochrome P450 CYP6AA3 from deltamethrin-selected resistant strain of Anopheles minimus mosquito, a major malaria vector in Thailand. In
Gold/platinum bimetallic core/shell nanoparticles stabilized by a Frechet-type dendrimer: preparation and catalytic hydrogenations of phenylaldehydes and nitrobenzenes.
Zhang W, et al.
Catalysis Letters, 127(3-4), 429-436 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service