Skip to Content
Merck
All Photos(1)

Documents

190284

Sigma-Aldrich

3-Phenoxybenzyl alcohol

98%

Synonym(s):

(3-Phenoxyphenyl)methanol, 1-Hydroxymethyl-3-phenoxybenzene, 3-(Hydroxymethyl)diphenyl ether, 3-Phenoxybenzenemethanol, m-Phenoxybenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4CH2OH
CAS Number:
Molecular Weight:
200.23
Beilstein:
475312
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.1 mmHg ( 37.7 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.591 (lit.)

bp

135-140 °C/0.1 mmHg (lit.)

density

1.149 g/mL at 25 °C (lit.)

SMILES string

OCc1cccc(Oc2ccccc2)c1

InChI

1S/C13H12O2/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-9,14H,10H2

InChI key

KGANAERDZBAECK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Phenoxybenzyl alcohol has been used in the synthesis of 3-phenoxylbenzyl β-D-glucuronide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

588.2 °F

Flash Point(C)

309 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Kaushik et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 36(3), 289-300 (2001-06-20)
The metabolism of fluvalinate in chickpea plants in the field under sub-tropical climatic conditions has been investigated. Ten days after spraying with fluvalinate, leaves were collected and extracted in acetone. The extract was fractionated and the metabolites were identified by
Development of a class selective immunoassay for the type II pyrethroid insecticides.
Mak SK, et al.
Analytica Chimica Acta, 534(1), 109-120 (2005)
G E Bast et al.
Archives of toxicology, 71(3), 179-186 (1997-01-01)
Isolated rabbit ears were single-pass perfused with a protein-free medium. Permethrin (0.05-23.5%, w/w) was applied in four distinct ointments. Permethrin, 3-phenoxybenzyl alcohol, 3-phenoxybenzaldehyde, and 3-phenoxybenzoic acid were analysed by HPLC. Permethrin was not detected in the effluent. The permeation coefficient
G E Bast et al.
Skin pharmacology and applied skin physiology, 11(4-5), 250-257 (1999-01-14)
The formation of 3-phenoxybenzoic acid from 3-phenoxybenzyl alcohol was determined in (a) rabbit ears, single-pass perfused with a protein-free buffer, pH 7.4; (b) the microsomal fraction and its supernatant from homogenized rabbit skin; and (c) purified alcohol dehydrogenase from horse
Sh T Iskandarova
Gigiena i sanitariia, (6)(6), 6-8 (1995-11-01)
The threshold level of 3-phenoxybenzyl alcohol (3-PBA) in ambient air was set up at 0.48 mg/m3. Inhalation of 3-PBA in concentrations of 4.86 and 0.518 mg/m3 caused changes in the central nervous system, metabolic processes, liver function, reduced the nonspecific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service