Skip to Content
Merck
All Photos(2)

Key Documents

186317

Sigma-Aldrich

N,N-Diethylformamide

99%

Synonym(s):

N-Formyldiethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(C2H5)2
CAS Number:
Molecular Weight:
101.15
Beilstein:
1209392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

176-177 °C (lit.)

density

0.908 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

CCN(CC)C=O

InChI

1S/C5H11NO/c1-3-6(4-2)5-7/h5H,3-4H2,1-2H3

InChI key

SUAKHGWARZSWIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Diethylformamide undergoes condensation with aromatic o-aminonitriles in the presence of ZnCl2 in sealed reactor to yield quinazoline-2,4(1H,3H)-dione.

Application

N,N-Diethylformamide was used in the synthesis of metal-organic frameworks. It was also used as solvent in the synthesis of porous cubic-shaped ZnO particles.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

South African research into AIDS "cure" severely criticised.
P Sidley
BMJ (Clinical research ed.), 314(7083), 771-771 (1997-03-15)
Jiarong Li et al.
Organic letters, 11(6), 1193-1196 (2009-02-26)
A new and facile preparation of quinazoline-2,4(1H,3H)-diones was first reported which was the condensation of aromatic o-aminonitriles with DMF or N,N-diethylformamide in the presence of ZnCl(2) (0.5-10 mol %) at 190-200 degrees C in the sealed reactor.
Yingmu Zhang et al.
Dalton transactions (Cambridge, England : 2003), 47(34), 11806-11811 (2018-06-05)
A new luminescent mesoporous metal-organic framework (PCN-604) was successfully synthesized from trimetric clusters, Al3(O)(CH3COO)6, and pyridine-based tritopic linkers, 4,4',4''-(pyridine-2,4,6-triyl)tribenzoate (PTB), via a solvothermal reaction. The MOF with an MTN topology possesses remarkable aqueous and thermal stabilities. Due to the efficient
Yuxia Shen et al.
Advanced materials (Deerfield Beach, Fla.), 30(52), e1802497-e1802497 (2018-11-06)
Large-scale synthesis of van der Waals (vdW) metal-organic framework (MOF) nanosheets with controlled crystallinity and interlayer coupling strength is one of the bottlenecks in 2D materials that has limited its successful transition to large-scale applications. Here, scalable synthesis of mBDC
Yu Kimitsuka et al.
Inorganic chemistry, 52(24), 14028-14033 (2013-11-26)
A method for synthesizing porous cubic-shaped ZnO particles a few tens of micrometers in size is described on the basis of a pyrolytic conversion of Zn-terephthalate metal-organic frameworks (MOFs). MOF crystals were initially grown in solutions containing Zn(NO3)2·6H2O and terephthalic

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service