Skip to Content
Merck
All Photos(2)

Key Documents

183075

Sigma-Aldrich

1-Methylisatin

97%

Synonym(s):

N-Methylindoline-2,3-dione, NSC 42449

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

130-133 °C (lit.)

SMILES string

CN1C(=O)C(=O)c2ccccc12

InChI

1S/C9H7NO2/c1-10-7-5-3-2-4-6(7)8(11)9(10)12/h2-5H,1H3

InChI key

VCYBVWFTGAZHGH-UHFFFAOYSA-N

Gene Information

human ... CASP3(836)

General description

The interaction between 1-methylisatin and human adult haemoglobin was studied using the circular dichroism (CD) spectroscopic, anisotropy and FTIR investigations.

Application

  • Reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones
  • Reactant for regioselective preparation of spirocyclic oxindole-butenolides
  • Reactant for synthesis of spiro-oxindoles
  • Reactant for preparation of unsymmetrical oxindoles
  • Reactant for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction
  • Reactant for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María C Rodríguez-Argüelles et al.
Journal of inorganic biochemistry, 101(1), 138-147 (2006-10-31)
Cobalt(II), nickel(II), copper(II) and zinc(II) complexes of 2-thiophenecarbonyl and isonicotinoyl hydrazones of 3-(N-methyl)isatin (HL(1) and HL(2), respectively) were synthesized and characterized, being the crystal structures of HL(1), HL(2) and [Ni(L(1))(2)].2CHCl(3) elucidated by X-ray diffraction techniques. The in vitro antimicrobial activity
Jaime F Martínez Suárez et al.
The Journal of organic chemistry, 84(11), 6879-6885 (2019-05-16)
Isatin in a solution of dry N,N-dimethylformamide/NaClO4 is electroreduced in the presence of CH3I. N-methylisatin (NMI) is obtained in quantitative molar yield and high current efficiency by controlled potential electrolysis (CPE). NMI and N-methylisatoic anhydride are the reaction products when
L Sebastian et al.
Letters in applied microbiology, 55(3), 234-239 (2012-06-29)
To investigate the drug to drug interaction of N-methylisatin-β-thiosemicarbazone (MIBT) derivative (SCH16) with ribavirin, mycophenolic acid and pentoxifylline against Japanese encephalitis virus in vitro. Our earlier studies have reported significant antiviral activity of these compounds against Japanese encephalitis virus in
Verena Schulz et al.
Organic letters, 9(9), 1745-1748 (2007-03-31)
[reaction: see text] The in situ preparation of a sulfonium ylide reagent achieved the highly diastereoselective epoxidation of isatins, so that a new and straightforward access to biologically significant spiro-epoxyoxindoles is provided. The first investigations of an asymmetric version are
M S Mirrlees et al.
Drug design and discovery, 11(3), 223-230 (1994-04-01)
Alkaline hydrolysis of the title isatin 4 in unbuffered solution obeys a rate law such that kobs varies; is directly proportional to [OH-]2 at pH 10 changes smoothly to kobs varies; is directly proportional to [OG-] by pH 13. As

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service