Skip to Content
Merck
All Photos(2)

Key Documents

177717

Sigma-Aldrich

2,5-Dimethylfuran

99%

Synonym(s):

2-Methyl-5-methylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
Beilstein:
106449
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

vapor density

3.31 (vs air)

Assay

99%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

92-94 °C (lit.)

mp

−62 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)o1

InChI

1S/C6H8O/c1-5-3-4-6(2)7-5/h3-4H,1-2H3

InChI key

GSNUFIFRDBKVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethylfuran is a heterocyclic furan used as a building block in organic synthesis.

Application

2,5-Dimethylfuran can be used as:
  • A reactant to prepare exo-maleimide-dimethylfuran cyclo adduct, which is used in the synthesis of oligonucleotide conjugates.
  • An arynophile to synthesize 1,4-dimethyl-1,4-dihydro-1,4-epoxynaphthalene (fused benzene) by reacting with a mixture of neopentyl glycol and 2-hydroxyphenylboronic acid in the presence of a strong base.
  • A precursor to prepare p-xylene via Diels-Alder cycloaddition and subsequent dehydration reaction.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F

Flash Point(C)

1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

5-hydroxymethylfurfural hydrodeoxygenation to 2, 5-dimethylfuran in continuous-flow system over Ni on nitrogen-doped carbon
Francesco B et al.
Sustainable Chemistry and Pharmacy, 1, 9-9 (2020)
Sara M Aschmann et al.
The journal of physical chemistry. A, 118(2), 457-466 (2014-01-02)
Products of the gas-phase reactions of OH radicals with furan, furan-d4, 2- and 3-methylfuran, and 2,3- and 2,5-dimethylfuran have been investigated in the presence of NO using direct air sampling atmospheric pressure ionization tandem mass spectrometry (API-MS and API-MS/MS), and
Production of renewable p-xylene from 2, 5-dimethylfuran via Diels-Alder cycloaddition and dehydrative aromatization reactions over silica- alumina aerogel catalysts
Wijaya YP, et al.
Catalysis Communications, 70, 12-16 (2015)
One-flow syntheses of diverse heterocyclic furan chemicals directly from fructose via tandem transformation platform
Guan-Young J et al.
NPG Asia materials, 7, e173-e173 (2015)
Simone M Charles et al.
Environmental science & technology, 42(4), 1324-1331 (2008-03-21)
Emissions of particulate matter (PM) and a broad suite of target volatile organic compounds (VOCs) in total, main-stream (MS) and side-stream (SS) smoke emissions are measured for six types of commercial cigarettes. The suitability of 2,5-dimethyl furan (DMF) as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service