All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4NNaO2
CAS Number:
Molecular Weight:
145.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
SMILES string
[Na+].[O-]C(=O)c1cccnc1
InChI
1S/C6H5NO2.Na/c8-6(9)5-2-1-3-7-4-5;/h1-4H,(H,8,9);/q;+1/p-1
InChI key
KFLRWGSAMLBHBV-UHFFFAOYSA-M
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wanda C Lakey et al.
Journal of clinical lipidology, 7(2), 178-181 (2013-02-19)
Multiple cholesterol-reducing therapies have been shown to induce the regression of tendon xanthoma in patients with familial hypercholesterolemia. We present 3 cases of adverse reactions in Achilles tendon xanthomas after the addition of niacin and bile acid sequestrants to ongoing
Rachel L Edwards et al.
Infection and immunity, 81(3), 945-955 (2013-01-16)
In response to environmental fluctuations or stresses, bacteria can activate transcriptional and phenotypic programs to coordinate an adaptive response. The intracellular pathogen Legionella pneumophila converts from a noninfectious replicative form to an infectious transmissive form when the bacterium encounters alterations
Alberto Zambon et al.
The American journal of cardiology, 113(9), 1494-1498 (2014-03-20)
Two large studies in populations selected for cardiovascular disease (CVD) demonstrated that raising high-density lipoprotein (HDL) cholesterol with niacin added to statin therapy did not decrease CVD. We examine the association of lipoprotein subfractions with niacin and changes in coronary
Malte Lohölter et al.
Archives of animal nutrition, 67(3), 202-218 (2013-06-08)
The objective of this study was to investigate the effects of niacin and dietary concentrate proportion on body temperature, ruminal pH and milk production of dairy cows. In a 2 × 2 factorial design, 20 primiparous Holstein cows (179 ±
Hyunju Shin et al.
Food chemistry, 138(2-3), 1109-1115 (2013-02-16)
An isotope dilution LC/mass spectrometric (ID-LC/MS) method was developed as a candidate reference method for the accurate determination of niacin in infant formula, breakfast cereals and multivitamin. After spiking nicotinamide-d(4) as an internal standard, infant formula and breakfast cereal samples
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service