Skip to Content
Merck
All Photos(4)

Documents

I17508

Sigma-Aldrich

Isonicotinic acid

99%

Synonym(s):

4-Picolinic acid, Pyridine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109599
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

technique(s)

cell culture | plant: suitable

mp

≥300 °C (lit.)

SMILES string

OC(=O)c1ccncc1

InChI

1S/C6H5NO2/c8-6(9)5-1-3-7-4-2-5/h1-4H,(H,8,9)

InChI key

TWBYWOBDOCUKOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isonicotinic acid (IN) can be used as:
  • An organocatalyst in the one pot four component condensation reaction, to synthesize pyranopyrazoles based heterocyclic compounds.
  • An organic ligand for the preparation of copper(I) halide coordination polymer [CuBr(IN)]n, by hydrothermal method.
  • As a starting material for the synthesis of cation-dimers with potent antimalarial activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimalarial Cation-dimers Synthesized in Two Steps from an Inexpensive Starting Material, Isonicotinic Acid
Motoshima K, et al.
ChemMedChem, 2(10), 1527-1532 (2007)
Synthesis of pyranopyrazoles using isonicotinic acid as a dual and biological organocatalyst
Zolfigol MA, et al.
Royal Society of Chemistry Advances, 3(48), 25681-25685 (2013)
Hydrothermal synthesis and characterization of a copper (I) halide coordination polymer with isonicotinic acid (IN) ligand as a template possessing three-dimensional supramolecular network structure
Yu J-H, et al.
Inorganic Chemistry Communications, 5(8), 572-576 (2002)
Rie Makiura et al.
Dalton transactions (Cambridge, England : 2003), 44(34), 15279-15285 (2015-06-03)
Surface modification of inorganic objects with metal-organic frameworks (MOFs) - organic-inorganic hybrid framework materials with infinite networks - opens wide windows for potential applications. In order to derive a target property, the key is the ability to fine tune the
Serena Berardi et al.
ChemSusChem, 8(21), 3688-3696 (2015-10-02)
Mastering the light-induced four-electron oxidation of water to molecular oxygen is a key step towards the achievement of overall water splitting to produce alternative solar fuels. In this work, we report two rugged molecular pyrazolate-based diruthenium complexes that efficiently catalyze

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service