Skip to Content
Merck
All Photos(1)

Key Documents

163821

Sigma-Aldrich

3-Methoxy-2-nitrobenzaldehyde

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CHO
CAS Number:
Molecular Weight:
181.15
Beilstein:
1959385
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97%

form

solid

mp

97-101 °C (lit.)

functional group

aldehyde
nitro

SMILES string

[H]C(=O)c1cccc(OC)c1[N+]([O-])=O

InChI

1S/C8H7NO4/c1-13-7-4-2-3-6(5-10)8(7)9(11)12/h2-5H,1H3

InChI key

GDTUACILWWLIJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxy-2-nitrobenzaldehyde undergoes 1,4-diazabicyclo[2.2.2]octane-catalyzed reaction with methyl vinyl ketone (MVK) to afford normal Baylis-Hillman adduct, the MVK dimer and a pair of diastereomeric bis-(MVK)Baylis-Hillman adducts[1].

Application

3-Methoxy-2-nitrobenzaldehyde was used in the synthesis of 8-hydroxyquinazoline, methy-3-methoxyanthranilate and 3-methoxy-2-nitrobenzylidenebisformamide[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

972. Analogues of 8-hydroxyquinoline having additional cyclic nitrogen atoms. Part I. Preparative.
Albert A and Hampton A.
Journal of the Chemical Society, 4985-4993 (1952)
Elucidation of the complex Baylis-Hillman reaction of 3-methoxy-2-nitrobenzaldehyde with methyl vinyl ketone.
Idahosa KC, et al.
South African Journal of Chemistry, 64, 144-150 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service