Skip to Content
Merck
All Photos(1)

Key Documents

156876

Sigma-Aldrich

N-(2-Hydroxyethyl)aniline

98%

Synonym(s):

2-(Phenylamino)ethanol, 2-Anilinoethanol, N-Phenylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH2CH2OH
CAS Number:
Molecular Weight:
137.18
Beilstein:
774672
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.578 (lit.)

bp

278-282 °C/760 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

OCCNc1ccccc1

InChI

1S/C8H11NO/c10-7-6-9-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2

InChI key

MWGATWIBSKHFMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-Hydroxyethyl)aniline was employed as substrate for human olfactory UDP-glucuronosyltransferase.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 - STOT SE 1

Target Organs

Blood, Blood,hematopoietic system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Santarem et al.
The Journal of organic chemistry, 73(16), 6466-6469 (2008-07-22)
An efficient formal total synthesis of (+)-gephyrotoxin is described. The key step of our strategy relies on the diastereoselective reduction of a chiral pyrrolidine beta-enamino ester obtained by condensation of ( S)-phenylglycinol on a protected 8-hydroxy-3,6-dioxooctanoate.
Katarina Babić et al.
Journal of chromatography. A, 1142(1), 84-92 (2006-10-19)
The performance of extractant impregnated resin (EIR) technology for chiral separation of amino-alcohols has been investigated. Phenylglycinol was selected as an archetype model enantiomer and azophenolic crown ether was used as a versatile enantioselective extractant. 1-Phenyloctane was selected as a
Alok K Awasthi et al.
The Journal of organic chemistry, 70(14), 5387-5397 (2005-07-02)
[reaction: see text] A practical, large-scale synthesis of a beta-amino ester 1 was developed. A chiral imine derived from (S)-phenylglycinol and 3-trimethylsilylpropanal was coupled with the Reformatsky reagent 3 with high diastereoselectivity (de > 98%) to give (SS)-4a as the
Mercedes Amat et al.
Chemical communications (Cambridge, England), (20)(20), 2935-2937 (2009-05-14)
The first total synthesis of (-)-16-episilicine has been completed from a phenylglycinol-derived bicyclic lactam, the key steps being stereoselective conjugate addition and alkylation reactions, and a ring-closing metathesis.
Mercedes Amat et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(28), 7724-7732 (2011-06-15)
Phenylglycinol-derived, unsaturated oxazolopiperidone lactams are extremely useful building blocks that undergo stereoselective conjugate addition reactions with organocuprates, enolates, and sulfur-stabilized anions, allowing the stereocontrolled introduction of substituents at the piperidine 4-position. The factors governing the exo- or endo-facial selectivity are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service