Skip to Content
Merck
All Photos(2)

Key Documents

156329

Sigma-Aldrich

Bisphenol A dimethacrylate

>98%

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane dimethacrylate, 2,2-Bis(4-methacryloxyphenyl)propane, 2,2-Bis(4-methacryloyloxyphenyl)propane, 4,4′-Isopropylidenediphenol dimethacrylate, BPADMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2C6H4]2C(CH3)2
CAS Number:
Molecular Weight:
364.43
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

>98%

form

solid

mp

72-74 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Oc1ccc(cc1)C(C)(C)c2ccc(OC(=O)C(C)=C)cc2

InChI

1S/C23H24O4/c1-15(2)21(24)26-19-11-7-17(8-12-19)23(5,6)18-9-13-20(14-10-18)27-22(25)16(3)4/h7-14H,1,3H2,2,4-6H3

InChI key

QUZSUMLPWDHKCJ-UHFFFAOYSA-N

General description

Bisphenol A dimethacrylate (BPA-DMA) is a key monomer, available in solid form with an assay of >98%, widely used in material science, particularly in the formulation of polymers and composite materials. BPA-DMA is primarily utilized as a crosslinking agent in the production of various thermosetting polymers. In dental applications, BPA-DMA is often incorporated into composites and adhesives due to its excellent bonding properties and ability to form durable, wear-resistant materials.

Application

Bisphenol A dimethacrylate (BPA-DMA) can be used as:
  • A template in the synthesis of molecularly imprinted polymers (MIPs). The use of BPA-DMA allows for the formation of specific cavities in the polymer matrix that match the shape and functional groups of BPA, enhancing the selectivity and affinity of the resulting polymer for its target analyte.
  • A co-monomer in the synthesis of photopolymerized monoliths for capillary electrochromatography.
  • A key component in the production of bisphenol A-glycidyl methacrylate (Bis-GMA), which is widely used in dental restorative materials due to its mechanical properties and potential for additional functionalities like antibacterial activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane C Atkinson et al.
Dental materials : official publication of the Academy of Dental Materials, 18(2), 128-135 (2002-01-05)
This study investigated the stability of compounds of dental sealant materials in a salivary matrix. Various amounts of bisphenol A (BPA), bisphenol A dimethacrylate (BIS-DMA) or triethylene-glycol dimethacrylate (TEGDMA) were added to whole salivary samples, and stored at -70 degrees
Faisal D Al-Qarni et al.
Journal of dentistry, 78, 91-99 (2018-08-29)
The objectives were to develop a calcium (Ca) and phosphate (P) ion-rechargeable and protein-repellent adhesive containing nanoparticles of amorphous calcium phosphate (NACP) and 2-methacryloyloxyethyl phosphorylcholine (MPC), and investigate the MPC effects on ion recharge and re-releases for the first time.
Alain Pefferkorn et al.
Dental materials : official publication of the Academy of Dental Materials, 24(12), 1647-1654 (2008-05-13)
The elaboration of efficient dental resins requires a high degree of internal cohesion and a strong organization of the filler/matrix systems, and a good compatibility between the organic and inorganic constituents of the composite. Combining fractal aerosils and polymer constitutes
G Eliades et al.
Dental materials : official publication of the Academy of Dental Materials, 17(4), 277-283 (2001-05-18)
The purpose of this study was to investigate whether monomer separation occurs in single-bottle adhesives applied on acid-etched dentin surfaces. Smear-layer covered dentin specimens produced from sound premolars, were acid-etched and analyzed by micro-MIR FTIR spectroscopy, before and after treatment
T E Schafer et al.
Journal of biomedical materials research, 45(3), 192-197 (1999-07-09)
Although pit and fissure sealants have been utilized extensively in dentistry as a way of preventing occlusal caries, results described by Olea et al. (1996) raised concerns about the safety of sealants and other resin-based dental materials due to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service