Skip to Content
Merck
All Photos(3)

Documents

151858

Sigma-Aldrich

Chloroform-d

"100%", 99.95 atom % D

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

isotopic purity

99.95 atom % D

Assay

≥99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform-d (Deuterochloroform, CDCl3), deuterated chloroform, is a 100% isotopically enriched NMR (Nuclear Magnetic Resonance) solvent. It is widely employed in high resolution NMR studies due to its high chemical and isotopic purity. Quantitaive infrared spectral investigations of carbon-deuterium stretching bands of chloroform-d in various organic solvents have been reported. Raman difference spectroscopic studies of mixtures of chloroform-d and liquid chloroform have been conducted to evaluate frequency shifts in the in the ν1 and ν2 bands of CHCl3 and CDCl3.

Application


  • Diphenylpyrrole-Strapped Calix[4]pyrrole Extractant for the Fluoride and Chloride Anions.: This study describes the development and application of a diphenylpyrrole-strapped calix[4]pyrrole as an effective extractant for fluoride and chloride anions, using Chloroform-d as a key solvent in its synthesis (Jung Heo N et al., 2023).

  • A simple protocol for determination of enantiopurity of amines using BINOL derivatives as chiral solvating agents via (1)H- and (19)F-NMR spectroscopic analysis.: This research outlines a protocol utilizing Chloroform-d for the NMR spectroscopic analysis of amines′ enantiopurity, highlighting its role in enhancing spectral quality and resolution (Chaudhary P et al., 2022).

  • Thermal oxidation stability of different multi-element oleogels via (1)H NMR spectroscopy.: Investigates the thermal oxidation stability of various oleogels, using Chloroform-d in the spectroscopic analysis to assess stability parameters (Qiu H et al., 2022).

  • Solvation of the Boc-Val-Phe-nPr peptide characterized by VCD spectroscopy and DFT calculations.: Explores solvation effects on a peptide′s structure, employing Chloroform-d as a solvent to determine its impact on peptide behavior in solution (Scholten K et al., 2022).

  • A straightforward catalytic approach to obtain deuterated chloroform at room temperature.: Discusses a novel method for producing Chloroform-d at room temperature, showcasing its practical applications in spectroscopic analysis and chemical synthesis (Higuera-Padilla AR et al., 2020).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daewoo Suh et al.
Nature communications, 11(1), 2252-2252 (2020-05-10)
Healable conductive materials have received considerable attention. However, their practical applications are impeded by low electrical conductivity and irreversible degradation after breaking/healing cycles. Here we report a highly conductive completely reversible electron tunneling-assisted percolation network of silver nanosatellite particles for
Zhenyang Si et al.
International archives of allergy and immunology, 182(10), 997-1007 (2021-08-25)
Cough-variant asthma (CVA) is a special type of asthma, solely manifesting with coughing. Studies suggest that airway inflammation is associated with CVA pathogenesis. Amygdalin is found to have an anti-inflammatory potential, while how it affects CVA remains unexplored. Cytotoxicity delivered
Isotopic dilution studies of the chloroform-chloroform-d system by Raman difference spectroscopy.
Laane J and Kiefer W.
J. Chem. Phys. , 73(10), 4971-4975 (1980)
Quantitative Study of the Bonding of Chloroform-d in Various Solvents by Infrared Spectrometry1.
Lord RC, et al.
Journal of the American Chemical Society, 77(5), 1365-1368 (1955)
Peng Zhang et al.
International journal of stem cells, 14(2), 229-239 (2021-02-27)
Implantation of bone marrow-derived mesenchymal stem cells (BMSCs) has been recognized as an effective therapy for attenuating acute lung injury (ALI). This study aims to discover microRNA (miRNA)-mediated improvement of BMSCs-based therapeutic effects. Mice were treated with lipopolysaccharide (LPS) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service