Skip to Content
Merck
All Photos(2)

Key Documents

144037

Sigma-Aldrich

1-Pyrenecarboxaldehyde

99%

Synonym(s):

1-Formylpyrene, 1-Pyrenealdehyde, 1-Pyrenecarbaldehyde, 3-Pyrenylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10O
CAS Number:
Molecular Weight:
230.26
Beilstein:
1874889
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

mp

123-126 °C (lit.)

SMILES string

[H]C(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C17H10O/c18-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12/h1-10H

InChI key

RCYFOPUXRMOLQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pyrenecarboxaldehyde (PCA) is a bifunctional molecule with pyrene moiety and one aldehyde group. The pyrene moiety interacts with the side walls of carbon nanotubes(CNT) through π-π stacking which leads to the uniform immobilization of PCA on the CNT surface. Therefore, it is widely used for the surface functionalization of CNTs. It can also be used to fabricate fluorescent chemosensors.

Features and Benefits

Exhibits reversible tuning properties with temperature and the properties depend upon the solvent environment.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tamara Zoltan et al.
Scientia pharmaceutica, 78(4), 767-789 (2010-12-24)
The synthesis of the meso-tetra(pyren-1-yl)porphyrin (1) was successfully accomplished by means of the pyrrole condensation with pyrene-1-carb-aldehyde in acidic media. Its metallization was carried out in an almost quantitative yield to obtain the corresponding complexes of Ni(II) (2), Cu(II) (3)
Yunyan Gao et al.
Talanta, 178, 663-669 (2017-11-16)
This work reports a facile strategy for the synthesis of water-soluble fluorescent probes Pyr1 and Pyr2, which have carboxyl and hydroxyl group in the side chain of thioacetal moiety, respectively. Pyr1-2 exhibit exclusively selective turn-on fluorescence response towards Hg
Aqueous Dispersion and Temperature Induced Reversible Fluorescence Properties of 1-Pyrenecarboxaldehyde
Ashaduzzaman, Md, and Masashi Kunitake
International Letters of Chemistry, Physics and Astronomy, 1, 55-62 (2013)
Jugun Prakash Chinta et al.
Langmuir : the ACS journal of surfaces and colloids, 33(22), 5519-5526 (2017-05-13)
Carbon nanotubes (CNTs) and semiconductor nanocrystals (SCNCs) are known to be interesting donor-acceptor partners due to their unique optical and electronic properties. These exciting features have led to the development of novel composites based on these two nanomaterials and to
Fluorescence probes for critical micelle concentration determination.
E D Goddard et al.
Langmuir : the ACS journal of surfaces and colloids, 1(3), 352-355 (1985-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service