Skip to Content
Merck
All Photos(4)

Documents

140155

Sigma-Aldrich

1,2-Dimethoxybenzene

ReagentPlus®, 99%

Synonym(s):

Pyrocatechol dimethyl ether, Veratrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Beilstein:
1364621
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.533 (lit.)

bp

206-207 °C (lit.)

mp

15 °C (lit.)

solubility

alcohol: soluble
diethyl ether: soluble
fatty oils: soluble
water: slightly soluble

density

1.084 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1OC

InChI

1S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6H,1-2H3

InChI key

ABDKAPXRBAPSQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dimethoxybenzene reacts with Li{N(SO2CF3)2} to yield molecular crystal [Li{N(SO2CF3)2}{C6H4(OCH3)2}2] having solid-state lithium ion conductivity. It is a potential pollinator attractant of the nocturnal moth Hadena bicruris.

Application

1,2-Dimethoxybenzene was used to investigate the electroantennogram response of vine weevil, Otiorhynchus sulcatus to plant volatiles.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tariq A Akhtar et al.
Plant physiology, 162(1), 52-62 (2013-04-03)
White campion (Silene latifolia) is a dioecious plant that emits 1,2-dimethoxybenzene (veratrole), a potent pollinator attractant to the nocturnal moth Hadena bicruris. Little is known about veratrole biosynthesis, although methylation of 2-methoxyphenol (guaiacol), another volatile emitted from white campion flowers
Makoto Moriya et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(40), 13554-13560 (2013-08-14)
The molecular crystals [Li{N(SO2CF3)2}{C6H4(OCH3)2}2] and [Li{N(SO2CF3)2}{C6F2H2(OCH3)2}2] with solid-state lithium ion conductivity have been synthesized by the addition of two equivalents of 1,2-dimethoxybenzene or 1,2-difluoro-4,5-dimethoxybenzene to Li{N(SO2CF3)2}, respectively. Single-crystal X-ray diffraction analysis revealed the formation of ionic conduction paths with an
Vincent J Chebny et al.
Organic letters, 11(11), 2253-2256 (2009-05-29)
Definitive X-ray crystallographic evidence is obtained for a single hole (or a polaron) to be uniformly distributed on the three equivalent 1,2-dimethoxybenzenoid (or veratrole) rings in the hexamethoxytriptycene cation radical. This conclusion is further supported by electrochemical analysis and by
Karsten Seidelmann et al.
Journal of insect physiology, 49(12), 1125-1133 (2003-11-20)
Mature gregarious male desert locusts, Schistocerca gregaria, emit the courtship inhibition pheromone phenylacetonitrile. Wings and legs, in particular the fore wings, have been identified as the main releasing sites. Abdomen and head emit only trace amounts of this pheromone. In
Olfactory antennal responses of the vine weevil Otiorhynchus sulcatus to plant volatiles.
Tol RWHM and Visser JH.
Entomologia Experimentalis et Applicata, 102(1), 49-64 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service