140066
1,3,5-Tribromobenzene
98%
Synonym(s):
1,3,5-Tribromobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
powder
bp
271 °C (lit.)
mp
117-121 °C (lit.)
SMILES string
Brc1cc(Br)cc(Br)c1
InChI
1S/C6H3Br3/c7-4-1-5(8)3-6(9)2-4/h1-3H
InChI key
YWDUZLFWHVQCHY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- 1,3,5-Tribromobenzene(TBB) acts as internal standard during derivatization step in determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry.
- TBB forms molecular complexes of 1:2 stoichiometry with [60]-and [70] fullerenes.
- TBB reacts with perfluoroalkenylzinc reagent in the presence of Pd(Ph3)4 catalyst to yield a novel trifunctional monomer 1,3,5-tris(α, β, β-trifluorovinyl)benzene.
Hazard Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(27), 3054-3058 (2009-09-08)
Cyanide (CN) is a powerful poison and rapidly toxic agent. Because of its wide availability and high toxicity, quantification of CN in blood and urine is frequently required in clinical and forensic practice. We present a sensitive and less time
Chemical communications (Cambridge, England), (20)(20), 2596-2597 (2003-11-05)
The reaction of the perfluoroalkenylzinc reagent, CF2=CFZnBr, with 1,3,5-tribromobenzene in the presence of a catalytic amount of Pd(Ph3)4 yielded a novel trifunctional monomer 1,3,5-tris(alpha,beta,beta-trifluorovinyl)benzene (1).
Journal of forensic sciences, 33(1), 217-222 (1988-01-01)
A sensitive analysis of sulfide in blood was established, using an extractive alkylation technique. Pentafluorobenzyl bromide was used as the alkylating agent, tetradecyldimethylbenzylammonium chloride as the phase-transfer catalyst, and potassium dihydrogenphosphate as the buffer to suppress the formation of sulfide.
Nucleic acids symposium series, (44)(44), 125-126 (2003-08-09)
We have designed a new type of a DNA dendrimer which has rigid branched structure. The branching molecule was prepared from 1,3,5-tribromobenzene. The dendrimer unit, in which three oligonucleotide-chains, two molecules of T15 and one molecule of A15, linked to
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(1-3), 66-70 (2005-11-01)
By UV-vis spectrophotometric method it has been shown that 1,3,5-tribromobenzene (TBB) forms molecular complexes of 1:2 stoichiometry with [60]- and [70]fullerenes. An isosbestic point could be detected in case of the [70]fullerene complex. The formation constant of the [60]fullerene complex
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service