Skip to Content
Merck
All Photos(2)

Key Documents

133035

Sigma-Aldrich

Methacrolein

95%

Synonym(s):

Methacrylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CHO
CAS Number:
Molecular Weight:
70.09
Beilstein:
1209258
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.4 (vs air)

vapor pressure

121 mmHg ( 20 °C)
13.44 psi ( 55 °C)
5.35 psi ( 20 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

68-70 °C (lit.)

mp

−81 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)C(C)=C

InChI

1S/C4H6O/c1-4(2)3-5/h3H,1H2,2H3

InChI key

STNJBCKSHOAVAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methacrolein on oxidation over heteropoly acid catalysts (mainly 12-molybdophosphoric acid) at 300°C yields methacrylic acid. It is the major product of isoprene oxidation.

Application

Methacrolein was used to study the effect of parts per billion levels of limonene oxidation products and the terpene oxidation product, methacrolein on human eye blink frequency. It was also used in determination of rate constants and the products of the reactions between atomic chlorine and acrolein, methacrolein and methyl vinyl ketone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S T Larsen et al.
Toxicology letters, 114(1-3), 197-202 (2000-03-14)
The acute respiratory effects of airborne exposure to methacrolein were studied in a recent refinement of the standard test method with mice (ASTM, 1984. American Society for Testing and Materials, Philadelphia). Irritation of the upper respiratory tract caused a concentration-dependent
Catalysis by heteropoly compounds: IV. Oxidation of methacrolein to methacrylic acid over 12-molybdophosphoric acid.
Konishi Y, et al.
J. Catal., 77(1), 169-179 (1982)
Michael N Paddon-Row et al.
The Journal of organic chemistry, 74(2), 861-868 (2008-12-11)
The cationic oxazaborolidine-catalyzed Diels-Alder reactions of butadiene and a series of five dienophiles have been studied using density functional theory (B3LYP/6-31G(d)). In each case, the computational results successfully reproduce the experimentally observed sense of stereoinduction and enantiomeric ratio. The computed
John D Crounse et al.
The journal of physical chemistry. A, 116(24), 5756-5762 (2012-03-29)
Peroxy radicals formed by addition of OH and O(2) to the olefinic carbon atoms in methacrolein react with NO to form methacrolein hydroxy nitrate and hydroxyacetone. We observe that the ratio of these two compounds, however, unexpectedly decreases as the
Henrik G Kjaergaard et al.
The journal of physical chemistry. A, 116(24), 5763-5768 (2012-03-29)
We investigate the oxidation of methacryloylperoxy nitrate (MPAN) and methacrylicperoxy acid (MPAA) by the hydroxyl radical (OH) theoretically, using both density functional theory [B3LYP] and explicitly correlated coupled cluster theory [CCSD(T)-F12]. These two compounds are produced following the abstraction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service