Skip to Content
Merck
All Photos(2)

Key Documents

131016

Sigma-Aldrich

Fumaronitrile

98%

Synonym(s):

trans-1,2-Dicyanoethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH=CHCN
CAS Number:
Molecular Weight:
78.07
Beilstein:
969245
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

186 °C (lit.)

mp

93-95 °C (lit.)

solubility

ethanol: 50 (mg/mL; colorless to yellow)

functional group

nitrile

SMILES string

N#C\C=C\C#N

InChI

1S/C4H2N2/c5-3-1-2-4-6/h1-2H/b2-1+

InChI key

KYPOHTVBFVELTG-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Crociani et al.
Dalton transactions (Cambridge, England : 2003), 41(40), 12490-12500 (2012-09-08)
The iminodiphosphine 2-(PPh(2))C(6)H(4)-1-CH=NC(6)H(4)-2-(PPh(2)) (P-N-P') is used for the preparation of the complexes [Pd(η(1)-CHR(1)-CH=CR(2)R(3))(P-N-P')]BF(4) [R(1) = R(2) = R(3) = H: (1); R(1) = R(2) = Ph, R(3) = H: (2); R(1) = R(3) = H, R(2) = Ph: (3); R(1)
Alicja B Veselá et al.
Applied microbiology and biotechnology, 100(5), 2193-2202 (2015-11-02)
The aim of this study was to discover new nitrilases with useful activities, especially towards dinitriles that are precursors of high-value cyano acids. Genes coding for putative nitrilases of different origins (fungal, plant, or bacterial) with moderate similarities to known
Stéphane Pagès et al.
The journal of physical chemistry. A, 110(24), 7547-7553 (2006-06-16)
The ultrafast ground state recovery (GSR) dynamics of the radical cation of perylene, Pe(*+), generated upon bimolecular photoinduced electron transfer in acetonitrile, has been investigated using pump-pump-probe spectroscopy. With 1,4-dicyanobenzene as electron acceptor, the free ion yield is substantial and
J M Wyatt et al.
Biodegradation, 6(2), 93-107 (1995-06-01)
Effluent from the manufacture of acrylonitrile is difficult to biodegrade. It contains nine major organic components: acetic acid, acrylonitrile, acrylamide, acrylic acid, acrolein, cyanopyridine, fumaronitrile, succinonitrile, and maleimide. A range of bacteria have been isolated that can grow on, or
R Schlegelmilch et al.
Journal of applied toxicology : JAT, 8(3), 201-209 (1988-06-01)
Two aliphatic nitriles, acetonitrile and fumaronitrile were tested for their genotoxic potential in three mutagenicity test systems: the Salmonella/microsome-assay, an assay using Saccharomyces cerevisiae (strain D7), and the bone marrow micronucleus test. Both compounds were tested with and without metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service