Skip to Content
Merck
All Photos(1)

Documents

127396

Sigma-Aldrich

Diphenyliodonium nitrate

Synonym(s):

DPIN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2INO3
CAS Number:
Molecular Weight:
343.12
Beilstein:
3922747
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

mp

150-154 °C (lit.)

λmax

203 nm
226 nm (2nd)

SMILES string

[O-][N+]([O-])=O.[I+](c1ccccc1)c2ccccc2

InChI

1S/C12H10I.NO3/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;2-1(3)4/h1-10H;/q+1;-1

InChI key

CQZCVYWWRJDZBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenyliodonium nitrate (DPIN) is a photoacid generator (PAG) that goes through photolysis reaction which can be used in the lithographic formation of UV reactive polymers. It can also be used in forming a light tuned self-assembled polyelectrolytic structures.

Application

Cationic photoinitiator. Photoacid generator.
DIPN can be used in the photo patterning of UV tuned shape memory hydrogels which can potentially be used in 3D printing.

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipeptide hydrogelation triggered via ultraviolet light
Raeburn J, et al.
Chemical Communications (Cambridge, England), 48(75), 9355-9357 (2012)
Alginate-graphene oxide hydrogels with enhanced ionic tunability and chemomechanical stability for light-directed 3D printing
Valentin TM, et al.
Carbon, 143, 447-456 (2019)
UV-controlled shape memory hydrogels triggered by photoacid generator
Feng W, et al.
Royal Society of Chemistry Advances, 5(100), 81784-81789 (2015)
Xingyi Li et al.
Briefings in bioinformatics, 21(2), 566-583 (2019-02-19)
Genes that are thought to be critical for the survival of organisms or cells are called essential genes. The prediction of essential genes and their products (essential proteins) is of great value in exploring the mechanism of complex diseases, the
Susana Castro et al.
Chemical communications (Cambridge, England), 48(72), 9089-9091 (2012-08-03)
A novel metal-free direct arylation of unbiased arenes with diaryliodonium triflates in a process where the use of solvents and additives is not required has been established.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service