Skip to Content
Merck
All Photos(2)

Documents

526940

Sigma-Aldrich

Triphenylsulfonium triflate

Synonym(s):

TPST, Triphenylsulfonium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3S(C6H5)3
CAS Number:
Molecular Weight:
412.45
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

mp

133-137 °C (lit.)

λmax

233 nm

SMILES string

[O-]S(=O)(=O)C(F)(F)F.c1ccc(cc1)[S+](c2ccccc2)c3ccccc3

InChI

1S/C18H15S.CHF3O3S/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1(3,4)8(5,6)7/h1-15H;(H,5,6,7)/q+1;/p-1

InChI key

FAYMLNNRGCYLSR-UHFFFAOYSA-M

General description

Triphenylsulfonium triflate (TPS-tf) is a photo-acid generator which can be incorporated in conjugated polymers to enhance the luminescence and reduce the operating voltage. The performance efficiency of the fabricated device is increased by improving the mobility of charge carriers.

Application

TPS-tfs can be used as photoacid generators for the preparation of a bilayer system based nanoporous template which can be potentially used in sensor applications. It can also be used to form a blend with polyfluorene-co-benzothiadiazole for the fabrication of organic light emitting diodes (OLEDs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of triphenylsulfonium triflate addition in wide band-gap polymer light-emitting diodes: improved charge injection, transport and electroplex-induced emission tuning.
Georgiadou DG, et al.
Royal Society of Chemistry Advances, 2(31), 11786-11792 (2012)
Mohammed S Almeataq et al.
Materials (Basel, Switzerland), 13(5) (2020-03-08)
Because of the increased demand for preceramic polymers in high-tech applications, there has been growing interest in the synthesis of preceramic polymers, including polysiloxanes and alumina. These polymers are preferred because of their low thermal expansion, conformability to surfaces over
Novel ArF excimer laser resists based on menthyl methacrylate terpolymer.
Shida N, et al.
J. Photopolym. Sci. Technol., 9(3), 457-464 (1996)
Fabrication of block copolymer templates by using dually responsive photoresist bottom layers.
Jung K, et al.
Reactive and Functional Polymers, 118(31), 20-25 (2017)
Shanhong Qi et al.
Die Pharmazie, 75(10), 505-509 (2020-12-12)
Objective: Intervertebral disc degeneration (IVDD) is very common in the elderly, so it is particularly important to find appropriate prevention or treatment. The aim of this study was to explore the effect of dexmedetomidine (DEX) on the degeneration of nucleus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service